Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462101_m1
          See other RALYL GE Assays ›
          Gene Symbol:
          Ralyl
          Gene Name:
          RALY RNA binding protein-like
          Gene Aliases:
          RGD1305844
          Chromosome Location:
          Chr.2: 89041591 - 89311720 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462101_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ralyl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024978.1 NP_001020149.1 4 - 5 400 111
          XM_006232123.2 XP_006232185.1 5 - 6 725 111
          XM_006232126.2 XP_006232188.1 5 - 6 1222 111
          XM_008760852.1 XP_008759074.1 5 - 6 725 111
          XM_008760853.1 XP_008759075.1 5 - 6 1222 111
          GenBank mRNA BC092614.1 - 4 - 5 400 7311942 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003238.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ralyl

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P236448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141757 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89248 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59079 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11259 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164271 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158653 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P236448 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P236448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P236448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822313 - -
          Affymetrix Rat Gene 1.0 ST Array 10822309 - -
          Affymetrix Rat Gene 1.0 ST Array 10822306 - -
          Affymetrix Rat Gene 1.0 ST Array 10822311 - -
          Affymetrix Rat Gene 1.0 ST Array 10822312 - -
          Affymetrix Rat Gene 1.1 ST Array 10822305 - -
          Rat Genome 230 2.0 Array 1380852_at - -
          Rat Genome 230 2.0 Array 1397513_PM_at - -
          Rat Genome 230 2.0 Array 1380852_PM_at - -
          Rat Genome 230 2.0 Array 1397513_at - -
          Affymetrix HT Rat Focus Array 1380852_PM_at - -
          Affymetrix HT Rat Focus Array 1380852_at - -
          Affymetrix Rat Clariom S Assay PSR0200022367.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022370.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022368.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022374.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022369.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022366.rn.1 TC0200003238.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200022373.rn.1 TC0200003238.rn.1 RTA TC
          Rat Expression Set 230 1397513_at - -
          Rat Expression Set 230 1380852_PM_at - -
          Rat Expression Set 230 1397513_PM_at - -
          Rat Expression Set 230 1380852_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822305 - -
          Rat Genome U34 Set rc_AI112346_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353565 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline