Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464727_m1
          See other MAPK9 GE Assays ›
          Gene Symbol:
          Mapk9
          Gene Name:
          mitogen-activated protein kinase 9
          Gene Aliases:
          SAPK
          Chromosome Location:
          Chr.10: 35333859 - 35375742 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464727_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270544.1 NP_001257473.1 9 - 10 1260 72
          NM_001270545.1 NP_001257474.1 9 - 10 1260 72
          NM_017322.2 NP_059018.1 9 - 10 1260 72
          XM_006246317.1 XP_006246379.1 9 - 10 1256 72
          XM_006246318.2 XP_006246380.1 9 - 10 1208 72
          GenBank mRNA BC061870.1 - 9 - 10 1227 5598943 72
          L27111.1 - 8 - 9 1016 72
          L27112.1 - 8 - 9 1016 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004343.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapk9

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          release of cytochrome c from mitochondria
          positive regulation of protein phosphorylation
          protein phosphorylation
          protein targeting to mitochondrion
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          JNK cascade
          JUN phosphorylation
          central nervous system development
          response to mechanical stimulus
          response to toxic substance
          response to organic substance
          positive regulation of gene expression
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cell morphogenesis involved in differentiation
          response to amine
          neuron projection development
          positive regulation of prostaglandin biosynthetic process
          regulation of protein ubiquitination
          positive regulation of prostaglandin secretion
          positive regulation of chemokine production
          cellular response to UV
          response to drug
          regulation of circadian rhythm
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription, DNA-templated
          regulation of JNK cascade
          response to cadmium ion
          rhythmic process
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          positive regulation of apoptotic signaling pathway
          JUN kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          mitogen-activated protein kinase kinase kinase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99850 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P480222 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P480222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P480222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217423 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451014 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700510453 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69074 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85983 - -
          Agilent Rat cDNA Microarray (G4105A) 700510453 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733166 - -
          Affymetrix Rat Gene 1.0 ST Array 10733175 - -
          Affymetrix Rat Gene 1.0 ST Array 10733173 - -
          Affymetrix Rat Gene 1.0 ST Array 10733165 - -
          Affymetrix Rat Gene 1.0 ST Array 10733161 - -
          Affymetrix Rat Gene 1.0 ST Array 10733170 - -
          Affymetrix Rat Gene 1.0 ST Array 10733171 - -
          Affymetrix Rat Gene 1.0 ST Array 10733168 - -
          Affymetrix Rat Gene 1.0 ST Array 10733176 - -
          Affymetrix Rat Gene 1.0 ST Array 10733162 - -
          Affymetrix Rat Gene 1.0 ST Array 10733163 - -
          Affymetrix Rat Gene 1.0 ST Array 10733172 - -
          Affymetrix Rat Gene 1.0 ST Array 10733177 - -
          Affymetrix Rat Gene 1.0 ST Array 10733167 - -
          Affymetrix Rat Gene 1.0 ST Array 10733164 - -
          Affymetrix Rat Gene 1.0 ST Array 10733174 - -
          Affymetrix Rat Gene 1.1 ST Array 10733160 - -
          Rat Genome 230 2.0 Array 1380024_at - -
          Rat Genome 230 2.0 Array 1378237_at - -
          Rat Genome 230 2.0 Array 1368646_PM_at - -
          Rat Genome 230 2.0 Array 1378237_PM_at - -
          Rat Genome 230 2.0 Array 1380024_PM_at - -
          Rat Genome 230 2.0 Array 1368646_at - -
          Affymetrix HT Rat Focus Array 1368646_at - -
          Affymetrix HT Rat Focus Array 1368646_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000040015.rn.1 TC1000004343.rn.1 RTA TC
          Rat Expression Set 230 1368646_at - -
          Rat Expression Set 230 1368646_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733160 - -
          Rat Genome U34 Set L27112_s_at - -
          Rat Genome U34 Set rc_AI231354_at - -
          Rat Genome U34 Set rc_AI231354_g_at - -
          Rat Genome U34 Set rc_AI231354_g_at - -
          Rat Genome U34 Set rc_AI231354_at - -
          Rat Genome U34 Set L27112_s_at - -
          Rat Genome U34 Set L27112_s_at - -
          Rat Genome U34 Set rc_AI231354_g_at - -
          Rat Genome U34 Set rc_AI231354_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357949 - -
          RatRef-12 Expression BeadChip ILMN_65467 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline