Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464736_g1
          See other CCL3 GE Assays ›
          Gene Symbol:
          Ccl3
          Gene Name:
          chemokine (C-C motif) ligand 3
          Gene Aliases:
          MIP-1a, Scya3
          Chromosome Location:
          Chr.10: 70869516 - 70871066 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464736_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013025.2 NP_037157.2 2 - 3 253 63
          GenBank mRNA EF121994.1 - 2 - 3 184 63
          EF121995.1 - 2 - 3 184 63
          U22414.1 - 2 - 3 253 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003430.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl3

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          osteoblast differentiation
          cell activation
          monocyte chemotaxis
          protein phosphorylation
          calcium ion transport
          cellular calcium ion homeostasis
          exocytosis
          chemotaxis
          inflammatory response
          cytoskeleton organization
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          regulation of cell shape
          response to toxic substance
          positive regulation of gene expression
          negative regulation of gene expression
          T cell chemotaxis
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          calcium-mediated signaling
          signaling
          positive regulation of cell migration
          negative regulation of bone mineralization
          neutrophil chemotaxis
          leukocyte chemotaxis
          lipopolysaccharide-mediated signaling pathway
          positive regulation of tumor necrosis factor production
          response to drug
          positive regulation of catalytic activity
          eosinophil degranulation
          protein kinase B signaling
          positive regulation of neuron apoptotic process
          astrocyte cell migration
          negative regulation by host of viral transcription
          negative regulation of osteoclast differentiation
          positive regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          eosinophil chemotaxis
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of interleukin-1 beta secretion
          positive regulation of inflammatory response
          positive regulation of calcium-mediated signaling
          positive chemotaxis
          positive regulation of protein kinase B signaling
          positive regulation of calcium ion transport
          regulation of sensory perception of pain
          cell chemotaxis
          positive regulation of ERK1 and ERK2 cascade
          response to cholesterol
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to organic cyclic compound
          granulocyte chemotaxis
          positive regulation of natural killer cell chemotaxis
          protein kinase activity
          calcium-dependent protein kinase C activity
          chemokine activity
          heparin binding
          phospholipase activator activity
          kinase activity
          CCR1 chemokine receptor binding
          CCR5 chemokine receptor binding
          chemoattractant activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P714311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P714311 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P714311 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P714311 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P714311 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10745682 - -
          Affymetrix Rat Gene 1.0 ST Array 10745681 - -
          Affymetrix Rat Gene 1.0 ST Array 10745679 - -
          Affymetrix Rat Gene 1.0 ST Array 10745680 - -
          Affymetrix Rat Gene 1.0 ST Array 10745678 - -
          Affymetrix Rat Gene 1.1 ST Array 10745677 - -
          Rat Genome 230 2.0 Array 1369815_at - -
          Rat Genome 230 2.0 Array 1369815_PM_at - -
          Affymetrix HT Rat Focus Array 1369815_PM_at - -
          Affymetrix HT Rat Focus Array 1369815_at - -
          Affymetrix Rat Clariom S Assay PSR1000030598.rn.1 TC1000003430.rn.1 RTA TC
          Rat Expression Set 230 1369815_PM_at - -
          Rat Expression Set 230 1369815_at - -
          Affymetrix Rat Gene 1.0 ST Array 10745677 - -
          Rat Genome U34 Set U22414_at - -
          Rat Genome U34 Set U22414_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22331826 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369286 - -
          RatRef-12 Expression BeadChip ILMN_54066 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline