Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01466738_m1
          See other MFAP3 GE Assays ›
          Gene Symbol:
          Mfap3
          Gene Name:
          microfibrillar-associated protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 42933077 - 42945591 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01466738_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfap3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007609.1 NP_001007610.1 2 - 3 468 143
          XM_006246379.2 XP_006246441.1 2 - 3 629 143
          GenBank mRNA BC079064.1 - 2 - 3 468 7121173 143
          EU000470.1 - 1 - 2 450 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000641.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mfap3

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P679269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P679269 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P679269 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P679269 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733727 - -
          Affymetrix Rat Gene 1.0 ST Array 10733728 - -
          Affymetrix Rat Gene 1.0 ST Array 10733729 - -
          Affymetrix Rat Gene 1.1 ST Array 10733726 - -
          Rat Genome 230 2.0 Array 1390063_PM_at - -
          Rat Genome 230 2.0 Array 1390063_at - -
          Affymetrix HT Rat Focus Array 1390063_at - -
          Affymetrix HT Rat Focus Array 1390063_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000005487.rn.1 TC1000000641.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000005488.rn.1 TC1000000641.rn.1 RTA TC
          Rat Expression Set 230 1390063_PM_at - -
          Rat Expression Set 230 1390063_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733726 - -
          Rat Genome U34 Set rc_AI639406_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351628 - -
          RatRef-12 Expression BeadChip ILMN_49527 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline