Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467656_g1
          See other NAA38 GE Assays ›
          Gene Symbol:
          Naa38
          Gene Name:
          N(alpha)-acetyltransferase 38, NatC auxiliary subunit
          Gene Aliases:
          Lsmd1, RGD1559617
          Chromosome Location:
          Chr.10: 55997508 - 55998691 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467656_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Naa38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105794.2 NP_001099264.2 2 - 3 549 96
          XM_008767795.1 XP_008766017.1 2 - 3 658 96
          GenBank mRNA FQ216987.1 - 2 - 3 319 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s)

          negative regulation of apoptotic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878885 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11203 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11203 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11203 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734971 - -
          Affymetrix Rat Gene 1.0 ST Array 10734974 - -
          Affymetrix Rat Gene 1.0 ST Array 10708153 - -
          Affymetrix Rat Gene 1.0 ST Array 10734973 - -
          Affymetrix Rat Gene 1.1 ST Array 10708152 - -
          Affymetrix Rat Gene 1.1 ST Array 10734970 - -
          Rat Genome 230 2.0 Array 1388683_PM_at - -
          Rat Genome 230 2.0 Array 1388683_at - -
          Affymetrix HT Rat Focus Array 1388683_at - -
          Affymetrix HT Rat Focus Array 1388683_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000007926.rn.1 TC1000000918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000007930.rn.1 TC1000000918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000007925.rn.1 TC1000000918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000007931.rn.1 TC1000000918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000007927.rn.1 TC1000000918.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000007933.rn.1 TC1000000918.rn.1 RTA TC
          Rat Expression Set 230 1388683_PM_at - -
          Rat Expression Set 230 1388683_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734970 - -
          Rat Genome U34 Set rc_AA859325_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21179011 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lksvr:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0