Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467675_g1
          See other CD300LF GE Assays ›
          Gene Symbol:
          Cd300lf
          Gene Name:
          Cd300 molecule-like family member F
          Gene Aliases:
          CD300f, RGD1309733
          Chromosome Location:
          Chr.10: 103669781 - 103690577 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467675_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd300lf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025111.1 NP_001020282.1 6 - 7 884 79
          XM_008768332.1 XP_008766554.1 7 - 8 1797 79
          XM_008768333.1 XP_008766555.1 8 - 9 1326 79
          XM_008768334.1 XP_008766556.1 7 - 8 1216 79
          XM_008768335.1 XP_008766557.1 6 - 7 1279 79
          XM_008768336.1 XP_008766558.1 6 - 7 1045 79
          XM_008768337.1 XP_008766559.1 7 - 8 1776 79
          XM_008768338.1 XP_008766560.1 7 - 8 1729 79
          XM_008768339.1 XP_008766561.1 7 - 8 1707 79
          XM_008768340.1 XP_008766562.1 6 - 7 1671 79
          XM_008768341.1 XP_008766563.1 7 - 8 1639 79
          XM_008768342.1 XP_008766564.1 6 - 7 1579 79
          GenBank mRNA BC093593.1 - 6 - 7 884 7314621 79
          GU057984.1 - 6 - 7 759 79
          GU062903.1 - 5 - 6 699 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004101.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd300lf

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s)

          osteoclast differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053675 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053675 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36000 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95329 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98648 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748879 - -
          Affymetrix Rat Gene 1.0 ST Array 10748875 - -
          Affymetrix Rat Gene 1.0 ST Array 10748874 - -
          Affymetrix Rat Gene 1.0 ST Array 10748878 - -
          Affymetrix Rat Gene 1.0 ST Array 10748877 - -
          Affymetrix Rat Gene 1.0 ST Array 10748880 - -
          Affymetrix Rat Gene 1.0 ST Array 10748872 - -
          Affymetrix Rat Gene 1.0 ST Array 10748876 - -
          Affymetrix Rat Gene 1.0 ST Array 10748881 - -
          Affymetrix Rat Gene 1.0 ST Array 10748873 - -
          Affymetrix Rat Gene 1.0 ST Array 10748871 - -
          Affymetrix Rat Gene 1.1 ST Array 10748870 - -
          Affymetrix Rat Clariom S Assay PSR1000037145.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037147.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037148.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037149.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037152.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000037146.rn.1 TC1000004101.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10748870 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21096156 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline