Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474592_m1
          See other UBXN7 GE Assays ›
          Gene Symbol:
          Ubxn7
          Gene Name:
          UBX domain protein 7
          Gene Aliases:
          RGD1306346, Ubxd7
          Chromosome Location:
          Chr.11: 71662786 - 71695031 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474592_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubxn7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107086.1 NP_001100556.1 1 - 2 127 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001755.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153340 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169626 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109442 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109442 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109442 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P942846 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P204246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754848 - -
          Affymetrix Rat Gene 1.0 ST Array 10754847 - -
          Affymetrix Rat Gene 1.0 ST Array 10754849 - -
          Affymetrix Rat Gene 1.1 ST Array 10754837 - -
          Rat Genome 230 2.0 Array 1392964_at - -
          Rat Genome 230 2.0 Array 1392964_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100011117.rn.1 TC1100001755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011118.rn.1 TC1100001755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011119.rn.1 TC1100001755.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011120.rn.1 TC1100001755.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10754837 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0