Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476495_m1
          See other MAP4K1 GE Assays ›
          Gene Symbol:
          Map4k1
          Gene Name:
          mitogen activated protein kinase kinase kinase kinase 1
          Gene Aliases:
          Hpk1
          Chromosome Location:
          Chr.1: 87937123 - 87959514 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476495_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map4k1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106243.1 NP_001099713.1 4 - 5 425 70
          XM_006228716.2 XP_006228778.1 4 - 5 1387 70
          XM_008759122.1 XP_008757344.1 4 - 5 1387 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001212.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKKK activity
          protein phosphorylation
          biological_process
          cell proliferation
          peptidyl-serine phosphorylation
          signal transduction by protein phosphorylation
          intracellular signal transduction
          protein autophosphorylation
          molecular_function
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          ATP binding
          MAP kinase kinase kinase kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P126085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60473 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32347 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P126085 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P126085 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P126085 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P126085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103153 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705700 - -
          Affymetrix Rat Gene 1.0 ST Array 10705701 - -
          Affymetrix Rat Gene 1.0 ST Array 10705695 - -
          Affymetrix Rat Gene 1.0 ST Array 10705686 - -
          Affymetrix Rat Gene 1.0 ST Array 10705685 - -
          Affymetrix Rat Gene 1.0 ST Array 10705707 - -
          Affymetrix Rat Gene 1.0 ST Array 10705699 - -
          Affymetrix Rat Gene 1.0 ST Array 10705696 - -
          Affymetrix Rat Gene 1.0 ST Array 10705687 - -
          Affymetrix Rat Gene 1.0 ST Array 10705706 - -
          Affymetrix Rat Gene 1.0 ST Array 10705694 - -
          Affymetrix Rat Gene 1.0 ST Array 10705703 - -
          Affymetrix Rat Gene 1.0 ST Array 10705702 - -
          Affymetrix Rat Gene 1.0 ST Array 10705708 - -
          Affymetrix Rat Gene 1.0 ST Array 10705691 - -
          Affymetrix Rat Gene 1.0 ST Array 10705684 - -
          Affymetrix Rat Gene 1.0 ST Array 10705681 - -
          Affymetrix Rat Gene 1.0 ST Array 10705689 - -
          Affymetrix Rat Gene 1.0 ST Array 10705710 - -
          Affymetrix Rat Gene 1.0 ST Array 10705711 - -
          Affymetrix Rat Gene 1.0 ST Array 10705697 - -
          Affymetrix Rat Gene 1.0 ST Array 10705692 - -
          Affymetrix Rat Gene 1.0 ST Array 10705680 - -
          Affymetrix Rat Gene 1.0 ST Array 10705690 - -
          Affymetrix Rat Gene 1.0 ST Array 10705709 - -
          Affymetrix Rat Gene 1.0 ST Array 10705688 - -
          Affymetrix Rat Gene 1.0 ST Array 10705682 - -
          Affymetrix Rat Gene 1.0 ST Array 10705705 - -
          Affymetrix Rat Gene 1.0 ST Array 10705698 - -
          Affymetrix Rat Gene 1.0 ST Array 10705683 - -
          Affymetrix Rat Gene 1.0 ST Array 10705693 - -
          Affymetrix Rat Gene 1.1 ST Array 10705679 - -
          Rat Genome 230 2.0 Array 1382803_at - -
          Rat Genome 230 2.0 Array 1376255_at - -
          Rat Genome 230 2.0 Array 1382803_PM_at - -
          Rat Genome 230 2.0 Array 1376255_PM_at - -
          Affymetrix HT Rat Focus Array 1382803_PM_at - -
          Affymetrix HT Rat Focus Array 1382803_at - -
          Affymetrix HT Rat Focus Array 1376255_at - -
          Affymetrix HT Rat Focus Array 1376255_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100009297.rn.1 TC0100001212.rn.1 RTA TC
          Rat Expression Set 230 1376255_at - -
          Rat Expression Set 230 1376255_PM_at - -
          Rat Expression Set 230 1382803_at - -
          Rat Expression Set 230 1382803_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705679 - -
          Rat Genome U34 Set rc_AA891302_at - -
          Rat Genome U34 Set rc_AA891302_g_at - -
          Rat Genome U34 Set rc_AI237423_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21331925 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371205 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0