Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477858_g1
          See other MRPL46 GE Assays ›
          Gene Symbol:
          Mrpl46
          Gene Name:
          mitochondrial ribosomal protein L46
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 140469557 - 140477634 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477858_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013068.1 NP_001013086.1 3 - 4 607 77
          GenBank mRNA BC086407.1 - 3 - 4 607 7306696 77
          FQ212933.1 - 3 - 4 603 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl46

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          structural constituent of ribosome
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P443008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106128 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P443008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P443008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P443008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P443008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10722889 - -
          Affymetrix Rat Gene 1.0 ST Array 10722886 - -
          Affymetrix Rat Gene 1.0 ST Array 10722888 - -
          Affymetrix Rat Gene 1.0 ST Array 10722887 - -
          Affymetrix Rat Gene 1.1 ST Array 10722885 - -
          Rat Genome 230 2.0 Array 1373155_PM_at - -
          Rat Genome 230 2.0 Array 1373155_at - -
          Affymetrix HT Rat Focus Array 1373155_PM_at - -
          Affymetrix HT Rat Focus Array 1373155_at - -
          Affymetrix Rat Clariom S Assay PSR0100046375.rn.1 TC0100005824.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100046374.rn.1 TC0100005824.rn.1 RTA TC
          Rat Expression Set 230 1373155_PM_at - -
          Rat Expression Set 230 1373155_at - -
          Affymetrix Rat Gene 1.0 ST Array 10722885 - -
          Rat Genome U34 Set rc_AI169228_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616392 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362149 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline