Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01480748_m1
          See other ORAI2 GE Assays ›
          Gene Symbol:
          Orai2
          Gene Name:
          ORAI calcium release-activated calcium modulator 2
          Gene Aliases:
          RGD1310213
          Chromosome Location:
          Chr.12: 23544483 - 23564602 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01480748_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Orai2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170403.1 NP_001163874.1 2 - 3 415 110
          GenBank mRNA BC158835.1 - 2 - 3 319 7384003 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000400.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452503 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019331 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019331 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P346822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93773 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757564 - -
          Affymetrix Rat Gene 1.0 ST Array 10757565 - -
          Affymetrix Rat Gene 1.0 ST Array 10757563 - -
          Affymetrix Rat Gene 1.1 ST Array 10757562 - -
          Rat Genome 230 2.0 Array 1379159_at - -
          Rat Genome 230 2.0 Array 1379159_PM_at - -
          Rat Genome 230 2.0 Array 1377955_PM_at - -
          Rat Genome 230 2.0 Array 1377955_at - -
          Affymetrix HT Rat Focus Array 1379159_PM_at - -
          Affymetrix HT Rat Focus Array 1377955_at - -
          Affymetrix HT Rat Focus Array 1377955_PM_at - -
          Affymetrix HT Rat Focus Array 1379159_at - -
          Affymetrix Rat Clariom S Assay PSR1200003555.rn.1 TC1200000400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003556.rn.1 TC1200000400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003551.rn.1 TC1200000400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003554.rn.1 TC1200000400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003557.rn.1 TC1200000400.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003558.rn.1 TC1200000400.rn.1 RTA TC
          Rat Expression Set 230 1377955_at - -
          Rat Expression Set 230 1379159_PM_at - -
          Rat Expression Set 230 1377955_PM_at - -
          Rat Expression Set 230 1379159_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757562 - -
          Rat Genome U34 Set rc_AI137602_at - -
          Rat Genome U34 Set rc_AI145132_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21889364 - -
          ABI Rat Genome Survey Microarray 22385465 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline