Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481987_m1
          See other PRMT5 GE Assays ›
          Gene Symbol:
          Prmt5
          Gene Name:
          protein arginine methyltransferase 5
          Gene Aliases:
          Skb1
          Chromosome Location:
          Chr.15: 33156508 - 33165893 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481987_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prmt5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108867.2 NP_001102337.2 16 - 17 1773 82
          GenBank mRNA BQ210423.1 - 1 - 2 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001670.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          spliceosomal snRNP assembly
          regulation of transcription, DNA-templated
          protein methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to symmetrical-dimethyl arginine
          circadian regulation of gene expression
          histone arginine methylation
          peptidyl-arginine N-methylation
          endothelial cell activation
          histone H4-R3 methylation
          regulation of DNA methylation
          negative regulation of cell differentiation
          positive regulation of oligodendrocyte differentiation
          regulation of ERK1 and ERK2 cascade
          liver regeneration
          core promoter sequence-specific DNA binding
          chromatin binding
          transcription corepressor activity
          methyltransferase activity
          histone-arginine N-methyltransferase activity
          protein complex binding
          protein-arginine omega-N symmetric methyltransferase activity
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P318846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P318846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P318846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P318846 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318843 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783567 - -
          Affymetrix Rat Gene 1.0 ST Array 10783560 - -
          Affymetrix Rat Gene 1.0 ST Array 10783571 - -
          Affymetrix Rat Gene 1.0 ST Array 10783555 - -
          Affymetrix Rat Gene 1.0 ST Array 10783568 - -
          Affymetrix Rat Gene 1.0 ST Array 10783565 - -
          Affymetrix Rat Gene 1.0 ST Array 10783554 - -
          Affymetrix Rat Gene 1.0 ST Array 10783559 - -
          Affymetrix Rat Gene 1.0 ST Array 10783557 - -
          Affymetrix Rat Gene 1.0 ST Array 10783556 - -
          Affymetrix Rat Gene 1.0 ST Array 10783558 - -
          Affymetrix Rat Gene 1.0 ST Array 10783569 - -
          Affymetrix Rat Gene 1.0 ST Array 10783564 - -
          Affymetrix Rat Gene 1.0 ST Array 10783562 - -
          Affymetrix Rat Gene 1.0 ST Array 10783561 - -
          Affymetrix Rat Gene 1.0 ST Array 10783566 - -
          Affymetrix Rat Gene 1.0 ST Array 10783563 - -
          Affymetrix Rat Gene 1.1 ST Array 10783553 - -
          Rat Genome 230 2.0 Array 1374424_at - -
          Rat Genome 230 2.0 Array 1374424_PM_at - -
          Affymetrix HT Rat Focus Array 1374424_at - -
          Affymetrix HT Rat Focus Array 1374424_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500010523.rn.1 TC1500001670.rn.1 RTA TC
          Rat Expression Set 230 1374424_PM_at - -
          Rat Expression Set 230 1374424_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783553 - -
          Rat Genome U34 Set rc_AI178944_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21687629 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350743 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline