Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01482475_m1
          See other MCF2L GE Assays ›
          Gene Symbol:
          Mcf2l
          Gene Name:
          MCF.2 cell line derived transforming sequence-like
          Gene Aliases:
          Ost, mcf 2l
          Chromosome Location:
          Chr.16: 81839691 - 81986671 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01482475_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcf2l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053951.2 NP_446403.2 5 - 6 509 69
          XM_006253416.2 XP_006253478.1 5 - 6 3771 69
          XM_006253419.2 XP_006253481.1 5 - 6 3771 69
          XM_006253421.2 XP_006253483.1 5 - 6 3770 69
          XM_006253423.2 XP_006253485.1 5 - 6 2397 69
          XM_006253424.2 XP_006253486.1 5 - 6 2588 69
          XM_008771381.1 XP_008769603.1 7 - 8 1739 69
          XM_008771382.1 XP_008769604.1 5 - 6 1141 69
          XM_008771383.1 XP_008769605.1 5 - 6 1424 69
          GenBank mRNA Z35654.1 - 3 - 4 605 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the Z35654.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mcf2l

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          Rho protein signal transduction
          intracellular signal transduction
          positive regulation of transcription from RNA polymerase II promoter
          Rho guanyl-nucleotide exchange factor activity
          1-phosphatidylinositol binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061786 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P531611 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P330812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125578 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164481 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4343 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P330812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P531611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061786 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P531611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P330812 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061786 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P330812 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061786 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P531611 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531611 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792852 - -
          Affymetrix Rat Gene 1.0 ST Array 10792856 - -
          Affymetrix Rat Gene 1.0 ST Array 10792846 - -
          Affymetrix Rat Gene 1.0 ST Array 10792853 - -
          Affymetrix Rat Gene 1.0 ST Array 10792847 - -
          Affymetrix Rat Gene 1.0 ST Array 10792845 - -
          Affymetrix Rat Gene 1.0 ST Array 10792854 - -
          Affymetrix Rat Gene 1.0 ST Array 10792849 - -
          Affymetrix Rat Gene 1.0 ST Array 10792851 - -
          Affymetrix Rat Gene 1.0 ST Array 10792855 - -
          Affymetrix Rat Gene 1.0 ST Array 10792848 - -
          Affymetrix Rat Gene 1.0 ST Array 10792857 - -
          Affymetrix Rat Gene 1.0 ST Array 10792844 - -
          Affymetrix Rat Gene 1.0 ST Array 10792850 - -
          Affymetrix Rat Gene 1.1 ST Array 10792843 - -
          Rat Genome 230 2.0 Array 1369820_PM_at - -
          Rat Genome 230 2.0 Array 1369820_at - -
          Affymetrix HT Rat Focus Array 1369820_PM_at - -
          Affymetrix HT Rat Focus Array 1369820_at - -
          Affymetrix Rat Clariom S Assay PSR1600015036.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015033.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015031.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015051.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015056.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015037.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015026.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015028.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015059.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015041.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015048.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015045.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015057.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015046.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015052.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015050.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015024.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015043.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015055.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015054.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015058.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015034.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015032.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015042.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015038.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015039.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015047.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015025.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015029.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015062.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015023.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015040.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015044.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015035.rn.1 TC1600002024.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600015027.rn.1 TC1600002024.rn.1 RTA TC
          Rat Expression Set 230 1369820_PM_at - -
          Rat Expression Set 230 1369820_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792843 - -
          Rat Genome U34 Set Z35654_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368963 - -
          RatRef-12 Expression BeadChip ILMN_54403 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline