Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483304_m1
          See other TMEM163 GE Assays ›
          Gene Symbol:
          Tmem163
          Gene Name:
          transmembrane protein 163
          Gene Aliases:
          RGD1306212, Sv31
          Chromosome Location:
          Chr.13: 44166306 - 44345735 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483304_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem163
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110763.2 NP_001104233.2 1 - 2 590 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001335.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43325 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57620 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30295 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121722 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95027 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111608 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119588 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119588 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119588 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P156647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61301 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28287 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83347 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767271 - -
          Affymetrix Rat Gene 1.0 ST Array 10767276 - -
          Affymetrix Rat Gene 1.0 ST Array 10767269 - -
          Affymetrix Rat Gene 1.0 ST Array 10767270 - -
          Affymetrix Rat Gene 1.0 ST Array 10767275 - -
          Affymetrix Rat Gene 1.0 ST Array 10767272 - -
          Affymetrix Rat Gene 1.0 ST Array 10767274 - -
          Affymetrix Rat Gene 1.0 ST Array 10767273 - -
          Affymetrix Rat Gene 1.1 ST Array 10767268 - -
          Rat Genome 230 2.0 Array 1375398_at - -
          Rat Genome 230 2.0 Array 1382222_PM_at - -
          Rat Genome 230 2.0 Array 1382222_at - -
          Rat Genome 230 2.0 Array 1375398_PM_at - -
          Affymetrix HT Rat Focus Array 1382222_PM_at - -
          Affymetrix HT Rat Focus Array 1382222_at - -
          Affymetrix HT Rat Focus Array 1375398_PM_at - -
          Affymetrix HT Rat Focus Array 1375398_at - -
          Affymetrix Rat Clariom S Assay PSR1300008708.rn.1 TC1300001335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008709.rn.1 TC1300001335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300008710.rn.1 TC1300001335.rn.1 RTA TC
          Rat Expression Set 230 1375398_at - -
          Rat Expression Set 230 1375398_PM_at - -
          Rat Expression Set 230 1382222_PM_at - -
          Rat Expression Set 230 1382222_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767268 - -
          Rat Genome U34 Set rc_AI058796_at - -
          Rat Genome U34 Set rc_AI007851_at - -
          Rat Genome U34 Set rc_AI234495_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21341826 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348880 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline