Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483598_m1
          See other FCGR3A GE Assays ›
          Gene Symbol:
          Fcgr3a
          Gene Name:
          Fc fragment of IgG, low affinity IIIa, receptor
          Gene Aliases:
          Fcgr4
          Chromosome Location:
          Chr.13: 89385773 - 89396047 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483598_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcgr3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207603.1 NP_997486.1 4 - 5 597 78
          XM_008769739.1 XP_008767961.1 5 - 6 682 78
          GenBank mRNA AY219230.1 - 4 - 5 597 78
          BC166877.1 - 4 - 5 624 6921627 78
          EF158003.1 - 4 - 5 628 78
          FQ225313.1 - 2 - 3 326 78
          FQ231465.1 - 2 - 3 326 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000748.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcgr3a

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          NK T cell proliferation
          immune response
          wound healing
          regulation of sensory perception of pain
          cellular response to lipopolysaccharide
          IgG binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P306439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P306439 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P306439 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306439 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73242 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765499 - -
          Affymetrix Rat Gene 1.0 ST Array 10765501 - -
          Affymetrix Rat Gene 1.0 ST Array 10765498 - -
          Affymetrix Rat Gene 1.0 ST Array 10765500 - -
          Affymetrix Rat Gene 1.0 ST Array 10765502 - -
          Affymetrix Rat Gene 1.1 ST Array 10765497 - -
          Rat Genome 230 2.0 Array 1373523_PM_at - -
          Rat Genome 230 2.0 Array 1373523_at - -
          Affymetrix HT Rat Focus Array 1373523_at - -
          Affymetrix HT Rat Focus Array 1373523_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300005160.rn.1 TC1300000748.rn.1 RTA TC
          Rat Expression Set 230 1373523_PM_at - -
          Rat Expression Set 230 1373523_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765497 - -
          Rat Genome U34 Set rc_AI011757_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21741051 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359086 - -
          RatRef-12 Expression BeadChip ILMN_55414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-snt5b:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline