Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01485112_m1
          See other MADD GE Assays ›
          Gene Symbol:
          Madd
          Gene Name:
          MAP-kinase activating death domain
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 79960301 - 80003023 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01485112_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Madd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053585.1 NP_446037.1 32 - 33 4849 83
          GenBank mRNA U72995.1 - 32 - 33 4849 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003648.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Madd

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          regulation of Rab protein signal transduction
          regulation of apoptotic process
          regulation of cell cycle
          negative regulation of growth hormone secretion
          execution phase of apoptosis
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of pancreatic amylase secretion
          negative regulation of apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway
          protein binding
          Rab guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P310089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76281 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46965 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101944 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P203195 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P203195 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P203195 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P531891 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P203195 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47718 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847343 - -
          Affymetrix Rat Gene 1.0 ST Array 10847326 - -
          Affymetrix Rat Gene 1.0 ST Array 10847328 - -
          Affymetrix Rat Gene 1.0 ST Array 10847319 - -
          Affymetrix Rat Gene 1.0 ST Array 10847346 - -
          Affymetrix Rat Gene 1.0 ST Array 10847337 - -
          Affymetrix Rat Gene 1.0 ST Array 10847334 - -
          Affymetrix Rat Gene 1.0 ST Array 10847313 - -
          Affymetrix Rat Gene 1.0 ST Array 10847331 - -
          Affymetrix Rat Gene 1.0 ST Array 10847314 - -
          Affymetrix Rat Gene 1.0 ST Array 10847320 - -
          Affymetrix Rat Gene 1.0 ST Array 10847321 - -
          Affymetrix Rat Gene 1.0 ST Array 10847329 - -
          Affymetrix Rat Gene 1.0 ST Array 10847311 - -
          Affymetrix Rat Gene 1.0 ST Array 10847317 - -
          Affymetrix Rat Gene 1.0 ST Array 10847323 - -
          Affymetrix Rat Gene 1.0 ST Array 10847340 - -
          Affymetrix Rat Gene 1.0 ST Array 10847347 - -
          Affymetrix Rat Gene 1.0 ST Array 10847324 - -
          Affymetrix Rat Gene 1.0 ST Array 10847339 - -
          Affymetrix Rat Gene 1.0 ST Array 10847342 - -
          Affymetrix Rat Gene 1.0 ST Array 10847345 - -
          Affymetrix Rat Gene 1.0 ST Array 10847344 - -
          Affymetrix Rat Gene 1.0 ST Array 10847322 - -
          Affymetrix Rat Gene 1.0 ST Array 10847336 - -
          Affymetrix Rat Gene 1.0 ST Array 10847309 - -
          Affymetrix Rat Gene 1.0 ST Array 10847348 - -
          Affymetrix Rat Gene 1.0 ST Array 10847325 - -
          Affymetrix Rat Gene 1.0 ST Array 10847312 - -
          Affymetrix Rat Gene 1.0 ST Array 10847330 - -
          Affymetrix Rat Gene 1.0 ST Array 10847333 - -
          Affymetrix Rat Gene 1.0 ST Array 10847316 - -
          Affymetrix Rat Gene 1.0 ST Array 10847349 - -
          Affymetrix Rat Gene 1.0 ST Array 10847338 - -
          Affymetrix Rat Gene 1.0 ST Array 10847350 - -
          Affymetrix Rat Gene 1.0 ST Array 10847335 - -
          Affymetrix Rat Gene 1.0 ST Array 10847310 - -
          Affymetrix Rat Gene 1.0 ST Array 10847341 - -
          Affymetrix Rat Gene 1.1 ST Array 10847308 - -
          Rat Genome 230 2.0 Array 1372580_PM_at - -
          Rat Genome 230 2.0 Array 1369066_PM_at - -
          Rat Genome 230 2.0 Array 1369066_at - -
          Rat Genome 230 2.0 Array 1372580_at - -
          Affymetrix HT Rat Focus Array 1369066_at - -
          Affymetrix HT Rat Focus Array 1372580_at - -
          Affymetrix HT Rat Focus Array 1369066_PM_at - -
          Affymetrix HT Rat Focus Array 1372580_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300027676.rn.1 TC0300003648.rn.1 RTA TC
          Rat Expression Set 230 1372580_PM_at - -
          Rat Expression Set 230 1369066_PM_at - -
          Rat Expression Set 230 1372580_at - -
          Rat Expression Set 230 1369066_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847308 - -
          Rat Genome U34 Set U72995_at - -
          Rat Genome U34 Set rc_AA848979_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21429428 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373958 - -
          RatRef-12 Expression BeadChip ILMN_65025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline