Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487769_m1
          See other PFKP GE Assays ›
          Gene Symbol:
          Pfkp
          Gene Name:
          phosphofructokinase, platelet
          Gene Aliases:
          ATP-PFK, PFK-C, PFK-P
          Chromosome Location:
          Chr.17: 68510761 - 68574447 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487769_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfkp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206847.1 NP_996729.1 20 - 21 2139 76
          XM_008771885.1 XP_008770107.1 20 - 21 2199 76
          GenBank mRNA BC088847.1 - 20 - 21 2139 7312293 76
          FN432820.1 - 20 - 21 2167 76
          L25387.1 - 20 - 21 2113 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002049.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfkp

          Panther Classification:

          Molecular Function -

          carbohydrate kinase kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          fructose 1,6-bisphosphate metabolic process
          carbohydrate phosphorylation
          protein homotetramerization
          6-phosphofructokinase activity
          ATP binding
          protein complex binding
          metal ion binding
          fructose-6-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95655 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99494 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014867 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014867 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014867 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15614 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700252592 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78022 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68270 - -
          Agilent Rat cDNA Microarray (G4105A) 700252592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796010 - -
          Affymetrix Rat Gene 1.0 ST Array 10795999 - -
          Affymetrix Rat Gene 1.0 ST Array 10795992 - -
          Affymetrix Rat Gene 1.0 ST Array 10796002 - -
          Affymetrix Rat Gene 1.0 ST Array 10796005 - -
          Affymetrix Rat Gene 1.0 ST Array 10796001 - -
          Affymetrix Rat Gene 1.0 ST Array 10795995 - -
          Affymetrix Rat Gene 1.0 ST Array 10796009 - -
          Affymetrix Rat Gene 1.0 ST Array 10795996 - -
          Affymetrix Rat Gene 1.0 ST Array 10796006 - -
          Affymetrix Rat Gene 1.0 ST Array 10795991 - -
          Affymetrix Rat Gene 1.0 ST Array 10795998 - -
          Affymetrix Rat Gene 1.0 ST Array 10796004 - -
          Affymetrix Rat Gene 1.0 ST Array 10795997 - -
          Affymetrix Rat Gene 1.0 ST Array 10796011 - -
          Affymetrix Rat Gene 1.0 ST Array 10796003 - -
          Affymetrix Rat Gene 1.0 ST Array 10795993 - -
          Affymetrix Rat Gene 1.0 ST Array 10795994 - -
          Affymetrix Rat Gene 1.0 ST Array 10796000 - -
          Affymetrix Rat Gene 1.0 ST Array 10796008 - -
          Affymetrix Rat Gene 1.0 ST Array 10796007 - -
          Affymetrix Rat Gene 1.0 ST Array 10795990 - -
          Affymetrix Rat Gene 1.1 ST Array 10795989 - -
          Rat Genome 230 2.0 Array 1372182_PM_at - -
          Rat Genome 230 2.0 Array 1372182_at - -
          Affymetrix HT Rat Focus Array 1372182_PM_at - -
          Affymetrix HT Rat Focus Array 1372182_at - -
          Affymetrix Rat Clariom S Assay PSR1700014043.rn.1 TC1700002049.rn.1 RTA TC
          Rat Expression Set 230 1372182_at - -
          Rat Expression Set 230 1372182_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795989 - -
          Rat Genome U34 Set L25387_g_at - -
          Rat Genome U34 Set L25387_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337118 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350318 - -
          RatRef-12 Expression BeadChip ILMN_49244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline