Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487945_m1
          See other LRP2BP GE Assays ›
          Gene Symbol:
          Lrp2bp
          Gene Name:
          Lrp2 binding protein
          Gene Aliases:
          RGD1305896
          Chromosome Location:
          Chr.16: 49437373 - 49463032 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrp2bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017443.1 NP_001017443.1 8 - 9 1167 71
          XM_006253141.2 XP_006253203.1 8 - 9 1083 71
          XM_006253143.2 XP_006253205.1 9 - 10 2164 71
          XM_008771257.1 XP_008769479.1 7 - 8 969 71
          GenBank mRNA BC092575.1 - 8 - 9 1167 7114719 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001656.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrp2bp

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P532115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65518 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P532115 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P532115 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P532115 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P532115 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47922 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24924 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19209 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791713 - -
          Affymetrix Rat Gene 1.0 ST Array 10791710 - -
          Affymetrix Rat Gene 1.0 ST Array 10791712 - -
          Affymetrix Rat Gene 1.0 ST Array 10791711 - -
          Affymetrix Rat Gene 1.0 ST Array 10791715 - -
          Affymetrix Rat Gene 1.0 ST Array 10791717 - -
          Affymetrix Rat Gene 1.0 ST Array 10791718 - -
          Affymetrix Rat Gene 1.0 ST Array 10791714 - -
          Affymetrix Rat Gene 1.0 ST Array 10791716 - -
          Affymetrix Rat Gene 1.1 ST Array 10791709 - -
          Affymetrix Rat Clariom S Assay PSR1600012364.rn.1 TC1600001656.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10791709 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21809791 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360899 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline