Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488384_m1
          See other CENPU GE Assays ›
          Gene Symbol:
          Cenpu
          Gene Name:
          centromere protein U
          Gene Aliases:
          Mlf1ip
          Chromosome Location:
          Chr.16: 48898032 - 48921248 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01488384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cenpu
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025673.1 NP_001020844.1 11 - 12 1187 115
          GenBank mRNA BC097399.1 - 11 - 12 1187 7453610 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001646.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cenpu

          Gene Ontology Categories:

          Function(s)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          chordate embryonic development

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P372810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372810 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P372810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44914 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96513 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791676 - -
          Affymetrix Rat Gene 1.0 ST Array 10791668 - -
          Affymetrix Rat Gene 1.0 ST Array 10791667 - -
          Affymetrix Rat Gene 1.0 ST Array 10791675 - -
          Affymetrix Rat Gene 1.0 ST Array 10791669 - -
          Affymetrix Rat Gene 1.0 ST Array 10791666 - -
          Affymetrix Rat Gene 1.0 ST Array 10791671 - -
          Affymetrix Rat Gene 1.0 ST Array 10791664 - -
          Affymetrix Rat Gene 1.0 ST Array 10791673 - -
          Affymetrix Rat Gene 1.0 ST Array 10791665 - -
          Affymetrix Rat Gene 1.0 ST Array 10791663 - -
          Affymetrix Rat Gene 1.0 ST Array 10791662 - -
          Affymetrix Rat Gene 1.0 ST Array 10791672 - -
          Affymetrix Rat Gene 1.0 ST Array 10791674 - -
          Affymetrix Rat Gene 1.0 ST Array 10791670 - -
          Affymetrix Rat Gene 1.1 ST Array 10791661 - -
          Rat Genome 230 2.0 Array 1392638_PM_at - -
          Rat Genome 230 2.0 Array 1393844_PM_at - -
          Rat Genome 230 2.0 Array 1392638_at - -
          Rat Genome 230 2.0 Array 1393844_at - -
          Affymetrix HT Rat Focus Array 1393844_at - -
          Affymetrix HT Rat Focus Array 1393844_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600012263.rn.1 TC1600001646.rn.1 RTA TC
          Rat Expression Set 230 1393844_PM_at - -
          Rat Expression Set 230 1393844_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791661 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371913 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline