Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488472_g1
          See other TLR3 GE Assays ›
          Gene Symbol:
          Tlr3
          Gene Name:
          toll-like receptor 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 50016468 - 50031011 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01488472_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tlr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198791.1 NP_942086.1 6 - 7 2862 142
          XM_008771266.1 XP_008769488.1 7 - 8 3111 142
          XM_008771267.1 XP_008769489.1 7 - 8 3031 142
          GenBank mRNA AB116229.1 - 6 - 7 2862 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000591.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tlr3

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          toll-like receptor signaling pathway
          microglial cell activation involved in immune response
          MyD88-independent toll-like receptor signaling pathway
          defense response
          I-kappaB phosphorylation
          male gonad development
          response to virus
          positive regulation of type I interferon production
          positive regulation of chemokine production
          positive regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor signaling pathway
          toll-like receptor 3 signaling pathway
          positive regulation of type III interferon production
          cellular response to interferon-beta
          cellular response to drug
          positive regulation of NF-kappaB import into nucleus
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          response to exogenous dsRNA
          response to dsRNA
          positive regulation of interferon-gamma biosynthetic process
          positive regulation of chemokine biosynthetic process
          positive regulation of interferon-alpha biosynthetic process
          positive regulation of interferon-beta biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          positive regulation of inflammatory response
          positive regulation of NF-kappaB transcription factor activity
          defense response to virus
          cellular response to mechanical stimulus
          cellular response to interferon-gamma
          cellular response to dsRNA
          cellular response to exogenous dsRNA
          extrinsic apoptotic signaling pathway
          necroptotic signaling pathway
          double-stranded RNA binding
          receptor activity
          transmembrane signaling receptor activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040806 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1021361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1021361 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040806 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1021361 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040806 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1021361 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1021361 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788241 - -
          Affymetrix Rat Gene 1.0 ST Array 10788245 - -
          Affymetrix Rat Gene 1.0 ST Array 10788239 - -
          Affymetrix Rat Gene 1.0 ST Array 10788240 - -
          Affymetrix Rat Gene 1.0 ST Array 10788244 - -
          Affymetrix Rat Gene 1.0 ST Array 10788243 - -
          Affymetrix Rat Gene 1.0 ST Array 10788242 - -
          Affymetrix Rat Gene 1.1 ST Array 10788238 - -
          Affymetrix Rat Clariom S Assay PSR1600004734.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004737.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004735.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004738.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004733.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600004736.rn.1 TC1600000591.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10788238 - -
          Rat Genome U34 Set rc_AA924130_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22178634 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline