Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01489555_m1
          See other COL4A3BP GE Assays ›
          Gene Symbol:
          Col4a3bp
          Gene Name:
          collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 27365148 - 27469797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01489555_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col4a3bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108935.1 NP_001102405.1 5 - 6 1021 82
          XM_006231827.2 XP_006231889.1 5 - 6 1017 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000223.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          in utero embryonic development
          heart morphogenesis
          ceramide metabolic process
          muscle contraction
          endoplasmic reticulum organization
          signal transduction
          cell proliferation
          phosphorylation
          response to endoplasmic reticulum stress
          ER to Golgi ceramide transport
          lipid homeostasis
          mitochondrion morphogenesis
          kinase activity
          ceramide transporter activity
          phosphatidylinositol-4-phosphate binding
          ceramide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036390 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036390 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036390 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P399459 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255449 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24935 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812626 - -
          Affymetrix Rat Gene 1.0 ST Array 10812616 - -
          Affymetrix Rat Gene 1.0 ST Array 10812618 - -
          Affymetrix Rat Gene 1.0 ST Array 10812622 - -
          Affymetrix Rat Gene 1.0 ST Array 10812613 - -
          Affymetrix Rat Gene 1.0 ST Array 10812615 - -
          Affymetrix Rat Gene 1.0 ST Array 10812617 - -
          Affymetrix Rat Gene 1.0 ST Array 10812619 - -
          Affymetrix Rat Gene 1.0 ST Array 10812628 - -
          Affymetrix Rat Gene 1.0 ST Array 10812614 - -
          Affymetrix Rat Gene 1.0 ST Array 10812624 - -
          Affymetrix Rat Gene 1.0 ST Array 10812629 - -
          Affymetrix Rat Gene 1.0 ST Array 10812611 - -
          Affymetrix Rat Gene 1.0 ST Array 10812612 - -
          Affymetrix Rat Gene 1.0 ST Array 10812620 - -
          Affymetrix Rat Gene 1.0 ST Array 10812625 - -
          Affymetrix Rat Gene 1.0 ST Array 10812627 - -
          Affymetrix Rat Gene 1.0 ST Array 10812621 - -
          Affymetrix Rat Gene 1.0 ST Array 10812623 - -
          Affymetrix Rat Gene 1.1 ST Array 10812610 - -
          Rat Genome 230 2.0 Array 1371893_at - -
          Rat Genome 230 2.0 Array 1371893_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200001182.rn.1 TC0200000223.rn.1 RTA TC
          Rat Expression Set 230 1371893_PM_at - -
          Rat Expression Set 230 1371893_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812610 - -
          Rat Genome U34 Set rc_AI112092_at - -
          Rat Genome U34 Set rc_AI228225_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22410097 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354291 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline