Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490170_m1
          See other SLC38A9 GE Assays ›
          Gene Symbol:
          Slc38a9
          Gene Name:
          solute carrier family 38, member 9
          Gene Aliases:
          RGD1311881
          Chromosome Location:
          Chr.2: 44512556 - 44592793 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490170_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc38a9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001035251.1 NP_001030328.1 11 - 12 1304 94
          XM_006231929.2 XP_006231991.1 12 - 13 1431 94
          XM_006231930.2 XP_006231992.1 13 - 14 1495 94
          XM_006231931.2 XP_006231993.1 12 - 13 1472 94
          XM_006231932.2 XP_006231994.1 12 - 13 1562 94
          XM_006231933.2 XP_006231995.1 12 - 13 1521 94
          XM_006231934.2 XP_006231996.1 11 - 12 3506 94
          XM_006231936.2 XP_006231998.1 11 - 12 3491 94
          XM_006231937.2 XP_006231999.1 11 - 12 1272 94
          XM_006231939.2 XP_006232001.1 11 - 12 1184 94
          GenBank mRNA BC092662.1 - 10 - 11 1166 94
          BC105869.1 - 11 - 12 1304 7442690 94
          JQ013729.1 - 11 - 12 1355 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000391.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc38a9

          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          sodium ion transport
          amino acid transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P316440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47330 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76805 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P316440 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P316440 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P316440 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P316440 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99808 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813038 - -
          Affymetrix Rat Gene 1.0 ST Array 10813041 - -
          Affymetrix Rat Gene 1.0 ST Array 10813030 - -
          Affymetrix Rat Gene 1.0 ST Array 10813034 - -
          Affymetrix Rat Gene 1.0 ST Array 10813036 - -
          Affymetrix Rat Gene 1.0 ST Array 10813032 - -
          Affymetrix Rat Gene 1.0 ST Array 10813037 - -
          Affymetrix Rat Gene 1.0 ST Array 10813031 - -
          Affymetrix Rat Gene 1.0 ST Array 10813035 - -
          Affymetrix Rat Gene 1.0 ST Array 10813043 - -
          Affymetrix Rat Gene 1.0 ST Array 10813039 - -
          Affymetrix Rat Gene 1.0 ST Array 10813042 - -
          Affymetrix Rat Gene 1.0 ST Array 10813033 - -
          Affymetrix Rat Gene 1.0 ST Array 10813040 - -
          Affymetrix Rat Gene 1.1 ST Array 10813029 - -
          Affymetrix Rat Clariom S Assay PSR0200002331.rn.1 TC0200000391.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10813029 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22380125 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364506 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline