Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490257_g1
          See other LOC498236 GE Assays ›
          Gene Symbol:
          LOC498236
          Gene Name:
          LRRGT00186
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 53030643 - 53087386 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490257_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC498236
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047926.1 NP_001041391.1 16 - 17 2094 81
          XM_008769565.1 XP_008767787.1 17 - 18 2293 81
          XM_008769566.1 XP_008767788.1 18 - 19 2299 81
          XM_008769567.1 XP_008767789.1 18 - 19 2299 81
          GenBank mRNA AY539937.1 - 16 - 17 2094 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001479.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P690802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109091 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P690802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P690802 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P690802 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P690802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70006 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768064 - -
          Affymetrix Rat Gene 1.0 ST Array 10768071 - -
          Affymetrix Rat Gene 1.0 ST Array 10768069 - -
          Affymetrix Rat Gene 1.0 ST Array 10768078 - -
          Affymetrix Rat Gene 1.0 ST Array 10768075 - -
          Affymetrix Rat Gene 1.0 ST Array 10768077 - -
          Affymetrix Rat Gene 1.0 ST Array 10768061 - -
          Affymetrix Rat Gene 1.0 ST Array 10768065 - -
          Affymetrix Rat Gene 1.0 ST Array 10768059 - -
          Affymetrix Rat Gene 1.0 ST Array 10768073 - -
          Affymetrix Rat Gene 1.0 ST Array 10768062 - -
          Affymetrix Rat Gene 1.0 ST Array 10768072 - -
          Affymetrix Rat Gene 1.0 ST Array 10768067 - -
          Affymetrix Rat Gene 1.0 ST Array 10768068 - -
          Affymetrix Rat Gene 1.0 ST Array 10768063 - -
          Affymetrix Rat Gene 1.0 ST Array 10768066 - -
          Affymetrix Rat Gene 1.0 ST Array 10768060 - -
          Affymetrix Rat Gene 1.0 ST Array 10768070 - -
          Affymetrix Rat Gene 1.0 ST Array 10768079 - -
          Affymetrix Rat Gene 1.0 ST Array 10768076 - -
          Affymetrix Rat Gene 1.0 ST Array 10768074 - -
          Affymetrix Rat Gene 1.1 ST Array 10768058 - -
          Rat Genome 230 2.0 Array 1391265_at - -
          Rat Genome 230 2.0 Array 1391265_PM_at - -
          Affymetrix HT Rat Focus Array 1391265_PM_at - -
          Affymetrix HT Rat Focus Array 1391265_at - -
          Affymetrix Rat Clariom S Assay PSR1300010183.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010200.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010202.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010204.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010184.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010192.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010197.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010198.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010207.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010209.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010215.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010187.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010217.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010182.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010214.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010218.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010190.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010199.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010210.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010211.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010216.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010193.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010196.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010185.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010186.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010194.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010203.rn.1 TC1300001479.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010205.rn.1 TC1300001479.rn.1 RTA TC
          Rat Expression Set 230 1391265_PM_at - -
          Rat Expression Set 230 1391265_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768058 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline