Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494192_m1
          See other POLR3G GE Assays ›
          Gene Symbol:
          Polr3g
          Gene Name:
          polymerase (RNA) III (DNA directed) polypeptide G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 9537642 - 9578551 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01494192_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr3g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109468.1 NP_001102938.1 6 - 7 1137 120
          XM_006231729.2 XP_006231791.1 5 - 6 479 120
          GenBank mRNA FQ209718.1 - 6 - 7 896 120
          FQ210837.1 - 6 - 7 896 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002528.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase III promoter
          biological_process
          cell proliferation
          positive regulation of interferon-beta production
          positive regulation of innate immune response
          RNA polymerase III activity
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089822 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123931 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089816 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123931 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089816 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123931 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089822 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123931 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089816 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78394 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820209 - -
          Affymetrix Rat Gene 1.0 ST Array 10820207 - -
          Affymetrix Rat Gene 1.0 ST Array 10820208 - -
          Affymetrix Rat Gene 1.0 ST Array 10718154 - -
          Affymetrix Rat Gene 1.0 ST Array 10718153 - -
          Affymetrix Rat Gene 1.0 ST Array 10820211 - -
          Affymetrix Rat Gene 1.0 ST Array 10820210 - -
          Affymetrix Rat Gene 1.1 ST Array 10718152 - -
          Affymetrix Rat Gene 1.1 ST Array 10820206 - -
          Rat Genome 230 2.0 Array 1382604_PM_at - -
          Rat Genome 230 2.0 Array 1382604_at - -
          Affymetrix Rat Clariom S Assay PSR0200017743.rn.1 TC0200002528.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017741.rn.1 TC0200002528.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017742.rn.1 TC0200002528.rn.1 RTA TC
          Rat Expression Set 230 1382604_at - -
          Rat Expression Set 230 1382604_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820206 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21564975 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline