Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498425_m1
          See other DBP GE Assays ›
          Gene Symbol:
          Dbp
          Gene Name:
          D site of albumin promoter (albumin D-box) binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 101687896 - 101692845 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498425_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289982.1 NP_001276911.1 3 - 4 856 72
          NM_001289983.1 NP_001276912.1 2 - 3 745 72
          NM_012543.3 NP_036675.1 3 - 4 1156 72
          GenBank mRNA AY518348.1 - 3 - 4 455 72
          AY518349.1 - 2 - 3 344 72
          BC087668.1 - 3 - 4 1156 7317132 72
          FQ213620.1 - 3 - 4 670 72
          J03179.1 - 3 - 4 1122 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001509.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbp

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          transcription, DNA-templated
          circadian rhythm
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017188 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049827 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P049828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P049828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049828 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P049827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017188 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P280898 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700765963 - -
          Agilent Rat cDNA Microarray (G4105A) 700765963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706958 - -
          Affymetrix Rat Gene 1.0 ST Array 10706961 - -
          Affymetrix Rat Gene 1.0 ST Array 10706956 - -
          Affymetrix Rat Gene 1.0 ST Array 10706954 - -
          Affymetrix Rat Gene 1.0 ST Array 10706955 - -
          Affymetrix Rat Gene 1.0 ST Array 10706959 - -
          Affymetrix Rat Gene 1.0 ST Array 10706957 - -
          Affymetrix Rat Gene 1.0 ST Array 10706960 - -
          Affymetrix Rat Gene 1.1 ST Array 10706953 - -
          Rat Genome 230 2.0 Array 1387874_at - -
          Rat Genome 230 2.0 Array 1387874_PM_at - -
          Affymetrix HT Rat Focus Array 1387874_at - -
          Affymetrix HT Rat Focus Array 1387874_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100012040.rn.1 TC0100001509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100012039.rn.1 TC0100001509.rn.1 RTA TC
          Rat Expression Set 230 1387874_at - -
          Rat Expression Set 230 1387874_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706953 - -
          Rat Genome U34 Set J03179_at - -
          Rat Genome U34 Set J03179_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21826972 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353765 - -
          RatRef-12 Expression BeadChip ILMN_51887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline