Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498801_m1
          See other MAP3K7CL GE Assays ›
          Gene Symbol:
          Map3k7cl
          Gene Name:
          MAP3K7 C-terminal like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 27208149 - 27251802 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01498801_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map3k7cl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013979.1 NP_001014001.1 4 - 5 524 62
          XM_006248016.2 XP_006248078.1 5 - 6 1236 62
          GenBank mRNA BC076389.1 - 4 - 5 524 7110014 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100000175.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Map3k7cl

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P532557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75979 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P532557 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P532557 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P532557 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P532557 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10750079 - -
          Affymetrix Rat Gene 1.0 ST Array 10750076 - -
          Affymetrix Rat Gene 1.0 ST Array 10750078 - -
          Affymetrix Rat Gene 1.0 ST Array 10750075 - -
          Affymetrix Rat Gene 1.0 ST Array 10750077 - -
          Affymetrix Rat Gene 1.1 ST Array 10750074 - -
          Rat Genome 230 2.0 Array 1384425_at - -
          Rat Genome 230 2.0 Array 1384425_PM_at - -
          Affymetrix HT Rat Focus Array 1384425_PM_at - -
          Affymetrix HT Rat Focus Array 1384425_at - -
          Affymetrix Rat Clariom S Assay PSR1100000658.rn.1 TC1100000175.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100000657.rn.1 TC1100000175.rn.1 RTA TC
          Rat Expression Set 230 1384425_PM_at - -
          Rat Expression Set 230 1384425_at - -
          Affymetrix Rat Gene 1.0 ST Array 10750074 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21924728 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360224 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline