Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499630_m1
          See other PRKAB1 GE Assays ›
          Gene Symbol:
          Prkab1
          Gene Name:
          protein kinase, AMP-activated, beta 1 non-catalytic subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 46316301 - 46326789 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499630_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkab1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031976.1 NP_114182.1 5 - 6 792 110
          XM_006249481.2 XP_006249543.1 6 - 7 792 110
          GenBank mRNA BC062008.1 - 5 - 6 678 5599996 110
          U42411.1 - 5 - 6 792 110
          X95577.1 - 6 - 7 762 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000759.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkab1

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          fatty acid biosynthetic process
          signal transduction
          positive regulation of gene expression
          regulation of protein kinase activity
          regulation of catalytic activity
          protein heterooligomerization
          protein kinase activity
          AMP-activated protein kinase activity
          protein binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1056161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177771 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056161 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056161 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1056161 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1056161 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702145885 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33815 - -
          Agilent Rat cDNA Microarray (G4105A) 702145885 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758942 - -
          Affymetrix Rat Gene 1.0 ST Array 10758946 - -
          Affymetrix Rat Gene 1.0 ST Array 10758943 - -
          Affymetrix Rat Gene 1.0 ST Array 10758944 - -
          Affymetrix Rat Gene 1.0 ST Array 10758945 - -
          Affymetrix Rat Gene 1.0 ST Array 10758948 - -
          Affymetrix Rat Gene 1.0 ST Array 10758947 - -
          Affymetrix Rat Gene 1.0 ST Array 10758941 - -
          Affymetrix Rat Gene 1.1 ST Array 10758940 - -
          Rat Genome 230 2.0 Array 1386945_a_at - -
          Rat Genome 230 2.0 Array 1386945_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386945_a_at - -
          Affymetrix HT Rat Focus Array 1386945_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1200006736.rn.1 TC1200000759.rn.1 RTA TC
          Rat Expression Set 230 1386945_a_at - -
          Rat Expression Set 230 1386945_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758940 - -
          Rat Genome U34 Set X95577_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21914479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline