Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01499727_m1
          See other GTF2I GE Assays ›
          Gene Symbol:
          Gtf2i
          Gene Name:
          general transcription factor II I
          Gene Aliases:
          Gtf2ird1
          Chromosome Location:
          Chr.12: 25410804 - 25487970 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01499727_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtf2i
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001512.2 NP_001001512.2 29 - 30 2843 140
          XM_006249162.2 XP_006249224.1 33 - 34 3108 140
          XM_006249163.2 XP_006249225.1 33 - 34 3105 140
          XM_006249164.2 XP_006249226.1 32 - 33 3048 140
          XM_006249165.2 XP_006249227.1 32 - 33 3045 140
          XM_006249166.2 XP_006249228.1 32 - 33 3042 140
          XM_006249167.2 XP_006249229.1 31 - 32 2991 140
          XM_006249168.2 XP_006249230.1 31 - 32 2985 140
          XM_006249169.2 XP_006249231.1 31 - 32 2970 140
          XM_006249170.2 XP_006249232.1 33 - 34 3303 140
          XM_006249171.2 XP_006249233.1 30 - 31 2928 140
          XM_006249172.2 XP_006249234.1 30 - 31 2913 140
          XM_006249173.2 XP_006249235.1 30 - 31 2907 140
          XM_006249174.2 XP_006249236.1 29 - 30 2850 140
          XM_008769141.1 XP_008767363.1 33 - 34 2970 140
          GenBank mRNA AF547390.1 - 31 - 32 2880 140
          AF547391.1 - 31 - 32 2880 140
          BC085815.1 - 29 - 30 2981 7123295 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000451.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gtf2i

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transition between slow and fast fiber
          negative regulation of angiogenesis
          negative regulation of cytosolic calcium ion concentration
          molecular_function
          DNA binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132841 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P110474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483662 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166987 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178057 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53479 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132841 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P110474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132841 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132841 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P110474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P120002 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8605 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5982 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25338 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72312 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16522 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757871 - -
          Affymetrix Rat Gene 1.0 ST Array 10757892 - -
          Affymetrix Rat Gene 1.0 ST Array 10757864 - -
          Affymetrix Rat Gene 1.0 ST Array 10757873 - -
          Affymetrix Rat Gene 1.0 ST Array 10757879 - -
          Affymetrix Rat Gene 1.0 ST Array 10757895 - -
          Affymetrix Rat Gene 1.0 ST Array 10757877 - -
          Affymetrix Rat Gene 1.0 ST Array 10757897 - -
          Affymetrix Rat Gene 1.0 ST Array 10757889 - -
          Affymetrix Rat Gene 1.0 ST Array 10757866 - -
          Affymetrix Rat Gene 1.0 ST Array 10757887 - -
          Affymetrix Rat Gene 1.0 ST Array 10757896 - -
          Affymetrix Rat Gene 1.0 ST Array 10757893 - -
          Affymetrix Rat Gene 1.0 ST Array 10757881 - -
          Affymetrix Rat Gene 1.0 ST Array 10757870 - -
          Affymetrix Rat Gene 1.0 ST Array 10757886 - -
          Affymetrix Rat Gene 1.0 ST Array 10757878 - -
          Affymetrix Rat Gene 1.0 ST Array 10757883 - -
          Affymetrix Rat Gene 1.0 ST Array 10757868 - -
          Affymetrix Rat Gene 1.0 ST Array 10757891 - -
          Affymetrix Rat Gene 1.0 ST Array 10757865 - -
          Affymetrix Rat Gene 1.0 ST Array 10757882 - -
          Affymetrix Rat Gene 1.0 ST Array 10757863 - -
          Affymetrix Rat Gene 1.0 ST Array 10757875 - -
          Affymetrix Rat Gene 1.0 ST Array 10757884 - -
          Affymetrix Rat Gene 1.0 ST Array 10757894 - -
          Affymetrix Rat Gene 1.0 ST Array 10757874 - -
          Affymetrix Rat Gene 1.0 ST Array 10757888 - -
          Affymetrix Rat Gene 1.0 ST Array 10757869 - -
          Affymetrix Rat Gene 1.0 ST Array 10757890 - -
          Affymetrix Rat Gene 1.0 ST Array 10757867 - -
          Affymetrix Rat Gene 1.0 ST Array 10757880 - -
          Affymetrix Rat Gene 1.0 ST Array 10757876 - -
          Affymetrix Rat Gene 1.0 ST Array 10757885 - -
          Affymetrix Rat Gene 1.1 ST Array 10757862 - -
          Rat Genome 230 2.0 Array 1385517_at - -
          Rat Genome 230 2.0 Array 1385517_PM_at - -
          Rat Genome 230 2.0 Array 1371931_at - -
          Rat Genome 230 2.0 Array 1371931_PM_at - -
          Affymetrix HT Rat Focus Array 1385517_PM_at - -
          Affymetrix HT Rat Focus Array 1385517_at - -
          Affymetrix HT Rat Focus Array 1371931_at - -
          Affymetrix HT Rat Focus Array 1371931_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200004120.rn.1 TC1200000451.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004119.rn.1 TC1200000451.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004121.rn.1 TC1200000451.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004122.rn.1 TC1200000451.rn.1 RTA TC
          Rat Expression Set 230 1371931_PM_at - -
          Rat Expression Set 230 1371931_at - -
          Rat Expression Set 230 1385517_PM_at - -
          Rat Expression Set 230 1385517_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757862 - -
          Rat Genome U34 Set rc_AI102849_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22335894 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline