Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507644_m1
          See other VKORC1 GE Assays ›
          Gene Symbol:
          Vkorc1
          Gene Name:
          vitamin K epoxide reductase complex, subunit 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 199338785 - 199341306 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507644_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vkorc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203335.2 NP_976080.1 2 - 3 309 94
          GenBank mRNA AY423047.1 - 2 - 3 292 94
          BC166413.1 - 305 9100951 94
          CB314647.1 - 2 - 3 325 94
          FQ210919.1 - 2 - 3 351 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006724.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vkorc1

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          blood coagulation
          response to organonitrogen compound
          response to organic cyclic compound
          drug metabolic process
          peptidyl-glutamic acid carboxylation
          regulation of blood coagulation
          vitamin K metabolic process
          response to antibiotic
          positive regulation of coagulation
          oxidation-reduction process
          bone development
          vitamin-K-epoxide reductase (warfarin-sensitive) activity
          vitamin-K-epoxide reductase (warfarin-insensitive) activity
          quinone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964982 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6734 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10257 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463345 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10726081 - -
          Affymetrix Rat Gene 1.0 ST Array 10726082 - -
          Affymetrix Rat Gene 1.0 ST Array 10726083 - -
          Affymetrix Rat Gene 1.0 ST Array 10726084 - -
          Affymetrix Rat Gene 1.1 ST Array 10726080 - -
          Rat Genome 230 2.0 Array 1388330_PM_at - -
          Rat Genome 230 2.0 Array 1388330_at - -
          Affymetrix HT Rat Focus Array 1388330_at - -
          Affymetrix HT Rat Focus Array 1388330_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100053055.rn.1 TC0100006724.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100053057.rn.1 TC0100006724.rn.1 RTA TC
          Rat Expression Set 230 1388330_at - -
          Rat Expression Set 230 1388330_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10726080 - -
          Rat Genome U34 Set rc_AI009197_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20846790 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355605 - -
          RatRef-12 Expression BeadChip ILMN_70001 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline