Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01511346_g1
          See other TGFB1I1 GE Assays ›
          Gene Symbol:
          Tgfb1i1
          Gene Name:
          transforming growth factor beta 1 induced transcript 1
          Gene Aliases:
          Ara55, Hic-5
          Chromosome Location:
          Chr.1: 199664112 - 199670970 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01511346_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb1i1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191840.1 NP_001178769.1 8 - 9 912 96
          XM_006230349.2 XP_006230411.1 8 - 9 1762 96
          XM_006230350.2 XP_006230412.1 8 - 9 884 96
          XM_008759920.1 XP_008758142.1 7 - 8 1969 96
          XM_008759921.1 XP_008758143.1 8 - 9 969 96
          GenBank mRNA AF314960.1 - 4 - 5 492 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002680.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to heat
          positive regulation of epithelial to mesenchymal transition
          Wnt signaling pathway
          morphogenesis of embryonic epithelium
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          ubiquitin-dependent SMAD protein catabolic process
          epithelial cell differentiation
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          transcription coactivator activity
          protein binding
          zinc ion binding
          Roundabout binding
          androgen receptor binding
          I-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P662897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P662897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P662897 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P662897 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P662897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29542 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711370 - -
          Affymetrix Rat Gene 1.0 ST Array 10711372 - -
          Affymetrix Rat Gene 1.0 ST Array 10711373 - -
          Affymetrix Rat Gene 1.0 ST Array 10711371 - -
          Affymetrix Rat Gene 1.0 ST Array 10711375 - -
          Affymetrix Rat Gene 1.0 ST Array 10711374 - -
          Affymetrix Rat Gene 1.1 ST Array 10711364 - -
          Rat Genome 230 2.0 Array 1370887_at - -
          Rat Genome 230 2.0 Array 1370887_PM_at - -
          Affymetrix HT Rat Focus Array 1370887_PM_at - -
          Affymetrix HT Rat Focus Array 1370887_at - -
          Affymetrix Rat Clariom S Assay PSR0100021005.rn.1 TC0100002680.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021007.rn.1 TC0100002680.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021004.rn.1 TC0100002680.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100021003.rn.1 TC0100002680.rn.1 RTA TC
          Rat Expression Set 230 1370887_PM_at - -
          Rat Expression Set 230 1370887_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711364 - -
          Rat Genome U34 Set rc_AA957362_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22011322 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355379 - -
          RatRef-12 Expression BeadChip ILMN_59428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline