Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01511827_m1
          See other GSTA3 GE Assays ›
          Gene Symbol:
          Gsta3
          Gene Name:
          glutathione S-transferase alpha 3
          Gene Aliases:
          Gsta5, Yc2
          Chromosome Location:
          Chr.9: 27357490 - 27417023 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01511827_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsta3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009920.2 NP_001009920.1 4 - 5 775 105
          NM_001159739.2 NP_001153211.1 5 - 6 745 105
          NM_001287025.1 NP_001273954.1 4 - 5 637 105
          GenBank mRNA BQ201014.1 - 1 - 2 105
          CO566642.1 - 5 - 6 608 105
          DY573045.1 - 3 - 4 470 105
          FQ210706.1 - 4 - 5 604 105
          FQ218526.1 - 5 - 6 618 105
          FQ219557.1 - 5 - 6 612 105
          FQ219615.1 - 5 - 6 745 105
          FQ219704.1 - 5 - 6 612 105
          S72506.1 - 4 - 5 623 105
          S82820.1 - 4 - 5 775 105
          X78847.1 - 4 - 5 623 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000352.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gsta3

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          aflatoxin catabolic process
          glutathione transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004231 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284349 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63207 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17332 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152026 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004231 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004231 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004231 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035291 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P491831 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701317988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14237 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6426 - -
          Agilent Rat cDNA Microarray (G4105A) 701317988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10922155 - -
          Affymetrix Rat Gene 1.0 ST Array 10922154 - -
          Affymetrix Rat Gene 1.0 ST Array 10922157 - -
          Affymetrix Rat Gene 1.0 ST Array 10922159 - -
          Affymetrix Rat Gene 1.0 ST Array 10922158 - -
          Affymetrix Rat Gene 1.0 ST Array 10922160 - -
          Affymetrix Rat Gene 1.0 ST Array 10922156 - -
          Affymetrix Rat Gene 1.0 ST Array 10922152 - -
          Affymetrix Rat Gene 1.1 ST Array 10922151 - -
          Rat Genome 230 2.0 Array 1371089_at - -
          Rat Genome 230 2.0 Array 1367774_at - -
          Rat Genome 230 2.0 Array 1371089_PM_at - -
          Rat Genome 230 2.0 Array 1367774_PM_at - -
          Rat Genome 230 2.0 Array 1368180_PM_s_at - -
          Rat Genome 230 2.0 Array 1368180_s_at - -
          Affymetrix HT Rat Focus Array 1371089_at - -
          Affymetrix HT Rat Focus Array 1371089_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900003027.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003032.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003028.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003026.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003030.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003025.rn.1 TC0900000352.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900003033.rn.1 TC0900000352.rn.1 RTA TC
          Rat Expression Set 230 1367774_at - -
          Rat Expression Set 230 1368180_s_at - -
          Rat Expression Set 230 1368180_PM_s_at - -
          Rat Expression Set 230 1367774_PM_at - -
          Rat Expression Set 230 1371089_PM_at - -
          Rat Expression Set 230 1371089_at - -
          Affymetrix Rat Gene 1.0 ST Array 10922151 - -
          Rat Genome U34 Set rc_AA945082_at - -
          Rat Genome U34 Set S82820mRNA_s_at - -
          Rat Genome U34 Set rc_AI235747_at - -
          Rat Genome U34 Set X78848cds_f_at - -
          Rat Genome U34 Set S72505_f_at - -
          Rat Genome U34 Set S72506_s_at - -
          Rat Genome U34 Set K01932_f_at - -
          Rat Genome U34 Set rc_AI235747_at - -
          Rat Genome U34 Set X78848cds_f_at - -
          Rat Genome U34 Set S72505_f_at - -
          Rat Genome U34 Set rc_AI235747_at - -
          Rat Genome U34 Set S82820mRNA_s_at - -
          Rat Genome U34 Set K01932_f_at - -
          Rat Genome U34 Set X78848cds_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21396897 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359094 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline