Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01513873_m1
          See other GALK2 GE Assays ›
          Gene Symbol:
          Galk2
          Gene Name:
          galactokinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 118144547 - 118268838 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01513873_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013919.1 NP_001013941.1 9 - 10 1246 66
          XM_006234904.2 XP_006234966.1 8 - 9 1701 66
          XM_006234906.2 XP_006234968.1 8 - 9 1206 66
          GenBank mRNA BC083716.1 - 9 - 10 1246 7190694 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001637.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galk2

          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          galactose metabolic process
          carbohydrate phosphorylation
          galactokinase activity
          ATP binding
          N-acetylgalactosamine kinase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P466523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42614 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43583 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60295 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P466523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P466523 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P466523 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P267444 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P466523 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63067 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84479 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97789 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839430 - -
          Affymetrix Rat Gene 1.0 ST Array 10839428 - -
          Affymetrix Rat Gene 1.0 ST Array 10839432 - -
          Affymetrix Rat Gene 1.0 ST Array 10839424 - -
          Affymetrix Rat Gene 1.0 ST Array 10839427 - -
          Affymetrix Rat Gene 1.0 ST Array 10839433 - -
          Affymetrix Rat Gene 1.0 ST Array 10839429 - -
          Affymetrix Rat Gene 1.0 ST Array 10839426 - -
          Affymetrix Rat Gene 1.0 ST Array 10839425 - -
          Affymetrix Rat Gene 1.0 ST Array 10839431 - -
          Affymetrix Rat Gene 1.1 ST Array 10839423 - -
          Rat Genome 230 2.0 Array 1372362_PM_at - -
          Rat Genome 230 2.0 Array 1372362_at - -
          Affymetrix HT Rat Focus Array 1372362_PM_at - -
          Affymetrix HT Rat Focus Array 1372362_at - -
          Affymetrix Rat Clariom S Assay PSR0300011490.rn.1 TC0300001637.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300011489.rn.1 TC0300001637.rn.1 RTA TC
          Rat Expression Set 230 1372362_PM_at - -
          Rat Expression Set 230 1372362_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839423 - -
          Rat Genome U34 Set rc_AI177657_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline