Search
Search
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007002.1 | NP_001007003.1 | 1 - 2 | 86 | 97 | |
| XM_006239574.1 | XP_006239636.1 | 1 - 2 | 454 | 97 | ||
| XM_006239575.1 | XP_006239637.1 | 1 - 2 | 454 | 88 | ||
| GenBank mRNA | BC083849.1 | - | 1 - 2 | 86 | 7122386 | 97 |
Set Membership: |
|