Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01519538_s1
          See other DDI2 GE Assays ›
          Gene Symbol:
          Ddi2
          Gene Name:
          DNA-damage inducible protein 2
          Gene Aliases:
          RGD1311815
          Chromosome Location:
          Chr.5: 160230958 - 160282835 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01519538_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddi2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034149.1 NP_001029321.1 10 3371 131
          XM_006239305.2 XP_006239367.1 9 3339 131
          XM_006239306.2 XP_006239368.1 9 3641 131
          XM_006239308.2 XP_006239370.1 2 4358 131
          XM_006239309.2 XP_006239371.1 10 3401 131
          GenBank mRNA BC097361.1 - 10 3371 7442573 131

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddi2

          Panther Classification:

          Molecular Function -

          aspartic protease membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          aspartic-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180828 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102708 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P201729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38699 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881177 - -
          Affymetrix Rat Gene 1.0 ST Array 10881175 - -
          Affymetrix Rat Gene 1.0 ST Array 10881171 - -
          Affymetrix Rat Gene 1.0 ST Array 10881170 - -
          Affymetrix Rat Gene 1.0 ST Array 10881168 - -
          Affymetrix Rat Gene 1.0 ST Array 10881176 - -
          Affymetrix Rat Gene 1.0 ST Array 10881169 - -
          Affymetrix Rat Gene 1.0 ST Array 10881173 - -
          Affymetrix Rat Gene 1.0 ST Array 10881172 - -
          Affymetrix Rat Gene 1.0 ST Array 10881174 - -
          Affymetrix Rat Gene 1.1 ST Array 10881167 - -
          Rat Genome 230 2.0 Array 1388609_at - -
          Rat Genome 230 2.0 Array 1388609_PM_at - -
          Rat Genome 230 2.0 Array 1379202_at - -
          Rat Genome 230 2.0 Array 1379202_PM_at - -
          Affymetrix HT Rat Focus Array 1388609_PM_at - -
          Affymetrix HT Rat Focus Array 1388609_at - -
          Rat Expression Set 230 1388609_at - -
          Rat Expression Set 230 1388609_PM_at - -
          Rat Expression Set 230 1379202_at - -
          Rat Expression Set 230 1379202_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881167 - -
          Rat Genome U34 Set rc_AI228230_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21228137 - -
          ABI Rat Genome Survey Microarray 21279859 - -
          ABI Rat Genome Survey Microarray 21529514 - -
          ABI Rat Genome Survey Microarray 22134037 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350060 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline