Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520350_m1
          See other MAN1C1 GE Assays ›
          Gene Symbol:
          Man1c1
          Gene Name:
          mannosidase, alpha, class 1C, member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 152775031 - 152920130 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Man1c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108687.1 NP_001102157.1 4 - 5 820 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003818.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          mannosyl-oligosaccharide 1,2-alpha-mannosidase activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029278 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773573 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139620 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169095 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13862 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1483 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57759 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12301 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029278 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029278 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029278 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P455038 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52671 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59175 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880551 - -
          Affymetrix Rat Gene 1.0 ST Array 10880546 - -
          Affymetrix Rat Gene 1.0 ST Array 10880547 - -
          Affymetrix Rat Gene 1.0 ST Array 10880548 - -
          Affymetrix Rat Gene 1.0 ST Array 10880540 - -
          Affymetrix Rat Gene 1.0 ST Array 10880541 - -
          Affymetrix Rat Gene 1.0 ST Array 10880550 - -
          Affymetrix Rat Gene 1.0 ST Array 10880544 - -
          Affymetrix Rat Gene 1.0 ST Array 10880543 - -
          Affymetrix Rat Gene 1.0 ST Array 10880545 - -
          Affymetrix Rat Gene 1.0 ST Array 10880549 - -
          Affymetrix Rat Gene 1.0 ST Array 10880542 - -
          Affymetrix Rat Gene 1.1 ST Array 10880539 - -
          Rat Genome 230 2.0 Array 1393053_at - -
          Rat Genome 230 2.0 Array 1398693_at - -
          Rat Genome 230 2.0 Array 1398693_PM_at - -
          Rat Genome 230 2.0 Array 1393053_PM_at - -
          Affymetrix HT Rat Focus Array 1393053_at - -
          Affymetrix HT Rat Focus Array 1393053_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500028668.rn.1 TC0500003818.rn.1 RTA TC
          Rat Expression Set 230 1393053_PM_at - -
          Rat Expression Set 230 1398693_PM_at - -
          Rat Expression Set 230 1398693_at - -
          Rat Expression Set 230 1393053_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880539 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22111039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline