Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522017_s1
          See other ADH6 GE Assays ›
          Gene Symbol:
          Adh6
          Gene Name:
          alcohol dehydrogenase 6 (class V)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 243656566 - 243687857 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522017_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012084.1 NP_001012084.1 8 1101 80
          XM_006233372.2 XP_006233434.1 8 1895 80
          XM_006233374.1 XP_006233436.1 9 1173 80
          XM_006233375.2 XP_006233437.1 10 1384 80
          XM_006233377.2 XP_006233439.1 8 1343 80
          XM_006233378.2 XP_006233440.1 9 1253 80
          XM_006233379.2 XP_006233441.1 9 1250 80
          XM_008761506.1 XP_008759728.1 7 1215 80
          XM_008761507.1 XP_008759729.1 9 3447 80
          XM_008761508.1 XP_008759730.1 10 3307 80
          XM_008761509.1 XP_008759731.1 10 1799 80
          XM_008761510.1 XP_008759732.1 10 3575 80
          GenBank mRNA BC083792.1 - 8 1101 7106499 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adh6

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          ethanol oxidation
          biological_process
          response to ethanol
          molecular_function
          alcohol dehydrogenase (NAD) activity
          alcohol dehydrogenase activity, zinc-dependent
          zinc ion binding
          ethanol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P199129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42260 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199129 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199129 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P199129 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49918 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67132 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56301 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819407 - -
          Affymetrix Rat Gene 1.0 ST Array 10819408 - -
          Affymetrix Rat Gene 1.0 ST Array 10819405 - -
          Affymetrix Rat Gene 1.0 ST Array 10819410 - -
          Affymetrix Rat Gene 1.0 ST Array 10819412 - -
          Affymetrix Rat Gene 1.0 ST Array 10819403 - -
          Affymetrix Rat Gene 1.0 ST Array 10819404 - -
          Affymetrix Rat Gene 1.0 ST Array 10819411 - -
          Affymetrix Rat Gene 1.0 ST Array 10819409 - -
          Affymetrix Rat Gene 1.1 ST Array 10819402 - -
          Rat Genome 230 2.0 Array 1395506_PM_at - -
          Rat Genome 230 2.0 Array 1395506_at - -
          Affymetrix Rat Clariom S Assay PSR0200016018.rn.1 TC0200002295.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016019.rn.1 TC0200002295.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016022.rn.1 TC0200002295.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016021.rn.1 TC0200002295.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10819402 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20723633 - -
          ABI Rat Genome Survey Microarray 21849871 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376848 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline