Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01522190_m1
          See other WDR63 GE Assays ›
          Gene Symbol:
          Wdr63
          Gene Name:
          WD repeat domain 63
          Gene Aliases:
          RGD1563105
          Chromosome Location:
          Chr.2: 251912432 - 251970699 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01522190_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134736.1 NP_001128208.1 18 - 19 2192 75
          GenBank mRNA BC168204.1 - 18 - 19 2192 7121248 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004861.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P393852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498763 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50268 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73030 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P393852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P393852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P393852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10827249 - -
          Affymetrix Rat Gene 1.0 ST Array 10827256 - -
          Affymetrix Rat Gene 1.0 ST Array 10827258 - -
          Affymetrix Rat Gene 1.0 ST Array 10827253 - -
          Affymetrix Rat Gene 1.0 ST Array 10827260 - -
          Affymetrix Rat Gene 1.0 ST Array 10827241 - -
          Affymetrix Rat Gene 1.0 ST Array 10827257 - -
          Affymetrix Rat Gene 1.0 ST Array 10827244 - -
          Affymetrix Rat Gene 1.0 ST Array 10827247 - -
          Affymetrix Rat Gene 1.0 ST Array 10827245 - -
          Affymetrix Rat Gene 1.0 ST Array 10827254 - -
          Affymetrix Rat Gene 1.0 ST Array 10827246 - -
          Affymetrix Rat Gene 1.0 ST Array 10827252 - -
          Affymetrix Rat Gene 1.0 ST Array 10827242 - -
          Affymetrix Rat Gene 1.0 ST Array 10827251 - -
          Affymetrix Rat Gene 1.0 ST Array 10827255 - -
          Affymetrix Rat Gene 1.0 ST Array 10827261 - -
          Affymetrix Rat Gene 1.0 ST Array 10827248 - -
          Affymetrix Rat Gene 1.0 ST Array 10827259 - -
          Affymetrix Rat Gene 1.0 ST Array 10827250 - -
          Affymetrix Rat Gene 1.0 ST Array 10827243 - -
          Affymetrix Rat Gene 1.1 ST Array 10827240 - -
          Affymetrix Rat Clariom S Assay PSR0200033635.rn.1 TC0200004861.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033640.rn.1 TC0200004861.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033634.rn.1 TC0200004861.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033637.rn.1 TC0200004861.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200033636.rn.1 TC0200004861.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10827240 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21343506 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline