Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528000_m1
          See other EMR4 GE Assays ›
          Gene Symbol:
          Emr4
          Gene Name:
          EGF-like module containing, mucin-like, hormone receptor-like sequence 4
          Gene Aliases:
          Emr4p
          Chromosome Location:
          Chr.9: 8056052 - 8199945 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528000_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emr4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007558.1 NP_001007559.1 1640 147
          XM_008766767.1 XP_008764989.1 1628 147
          GenBank mRNA AY686633.1 - 1640 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000065.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Emr4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021325 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47787 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103708 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10298 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97147 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86882 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18616 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88639 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67584 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40004 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9369 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91829 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10930718 - -
          Affymetrix Rat Gene 1.0 ST Array 10930722 - -
          Affymetrix Rat Gene 1.0 ST Array 10930729 - -
          Affymetrix Rat Gene 1.0 ST Array 10930721 - -
          Affymetrix Rat Gene 1.0 ST Array 10930725 - -
          Affymetrix Rat Gene 1.0 ST Array 10930724 - -
          Affymetrix Rat Gene 1.0 ST Array 10930720 - -
          Affymetrix Rat Gene 1.0 ST Array 10930719 - -
          Affymetrix Rat Gene 1.0 ST Array 10930723 - -
          Affymetrix Rat Gene 1.0 ST Array 10930727 - -
          Affymetrix Rat Gene 1.0 ST Array 10930732 - -
          Affymetrix Rat Gene 1.0 ST Array 10930726 - -
          Affymetrix Rat Gene 1.0 ST Array 10930728 - -
          Affymetrix Rat Gene 1.0 ST Array 10930730 - -
          Affymetrix Rat Gene 1.0 ST Array 10930731 - -
          Affymetrix Rat Gene 1.1 ST Array 10930717 - -
          Affymetrix Rat Clariom S Assay PSR0900000352.rn.1 TC0900000065.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900000353.rn.1 TC0900000065.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900000354.rn.1 TC0900000065.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10930717 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616521 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline