Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536269_m1
          See other MYH7B GE Assays ›
          Gene Symbol:
          Myh7b
          Gene Name:
          myosin, heavy chain 7B, cardiac muscle, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 151105043 - 151150621 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536269_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myh7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107794.2 NP_001101264.2 23 - 24 2835 111
          XM_008762338.1 XP_008760560.1 24 - 25 3696 111
          XM_008762339.1 XP_008760561.1 24 - 25 3102 111
          XM_008762340.1 XP_008760562.1 24 - 25 2839 111
          XM_008762341.1 XP_008760563.1 24 - 25 3696 111
          GenBank mRNA EU241478.1 - 23 - 24 2835 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002120.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          motor activity
          actin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P073976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44173 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151258 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073974 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P073974 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P073974 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208601 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65515 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34541 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841384 - -
          Affymetrix Rat Gene 1.0 ST Array 10841393 - -
          Affymetrix Rat Gene 1.0 ST Array 10841386 - -
          Affymetrix Rat Gene 1.0 ST Array 10841392 - -
          Affymetrix Rat Gene 1.0 ST Array 10841379 - -
          Affymetrix Rat Gene 1.0 ST Array 10841372 - -
          Affymetrix Rat Gene 1.0 ST Array 10841369 - -
          Affymetrix Rat Gene 1.0 ST Array 10841405 - -
          Affymetrix Rat Gene 1.0 ST Array 10841371 - -
          Affymetrix Rat Gene 1.0 ST Array 10841396 - -
          Affymetrix Rat Gene 1.0 ST Array 10841407 - -
          Affymetrix Rat Gene 1.0 ST Array 10841397 - -
          Affymetrix Rat Gene 1.0 ST Array 10841398 - -
          Affymetrix Rat Gene 1.0 ST Array 10841408 - -
          Affymetrix Rat Gene 1.0 ST Array 10841394 - -
          Affymetrix Rat Gene 1.0 ST Array 10841380 - -
          Affymetrix Rat Gene 1.0 ST Array 10841370 - -
          Affymetrix Rat Gene 1.0 ST Array 10841403 - -
          Affymetrix Rat Gene 1.0 ST Array 10841389 - -
          Affymetrix Rat Gene 1.0 ST Array 10841406 - -
          Affymetrix Rat Gene 1.0 ST Array 10841373 - -
          Affymetrix Rat Gene 1.0 ST Array 10841387 - -
          Affymetrix Rat Gene 1.0 ST Array 10841400 - -
          Affymetrix Rat Gene 1.0 ST Array 10841395 - -
          Affymetrix Rat Gene 1.0 ST Array 10841381 - -
          Affymetrix Rat Gene 1.0 ST Array 10841402 - -
          Affymetrix Rat Gene 1.0 ST Array 10841377 - -
          Affymetrix Rat Gene 1.0 ST Array 10841382 - -
          Affymetrix Rat Gene 1.0 ST Array 10841376 - -
          Affymetrix Rat Gene 1.0 ST Array 10841385 - -
          Affymetrix Rat Gene 1.0 ST Array 10841383 - -
          Affymetrix Rat Gene 1.0 ST Array 10841409 - -
          Affymetrix Rat Gene 1.0 ST Array 10841374 - -
          Affymetrix Rat Gene 1.0 ST Array 10841391 - -
          Affymetrix Rat Gene 1.0 ST Array 10841399 - -
          Affymetrix Rat Gene 1.0 ST Array 10841401 - -
          Affymetrix Rat Gene 1.0 ST Array 10841375 - -
          Affymetrix Rat Gene 1.0 ST Array 10841378 - -
          Affymetrix Rat Gene 1.0 ST Array 10841388 - -
          Affymetrix Rat Gene 1.0 ST Array 10841390 - -
          Affymetrix Rat Gene 1.0 ST Array 10841404 - -
          Affymetrix Rat Gene 1.1 ST Array 10841367 - -
          Rat Genome 230 2.0 Array 1371584_at - -
          Rat Genome 230 2.0 Array 1371584_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300015564.rn.1 TC0300002120.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10841367 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22290209 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365040 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline