Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536488_m1
          See other CDH16 GE Assays ›
          Gene Symbol:
          Cdh16
          Gene Name:
          cadherin 16
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 568329 - 578513 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536488_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdh16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012055.1 NP_001012055.1 17 - 18 2538 89
          GenBank mRNA BC081996.1 - 17 - 18 2538 7130691 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000015.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdh16

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544249 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052765 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052765 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P302334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16952 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26778 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805668 - -
          Affymetrix Rat Gene 1.0 ST Array 10805662 - -
          Affymetrix Rat Gene 1.0 ST Array 10805669 - -
          Affymetrix Rat Gene 1.0 ST Array 10805667 - -
          Affymetrix Rat Gene 1.0 ST Array 10805658 - -
          Affymetrix Rat Gene 1.0 ST Array 10805665 - -
          Affymetrix Rat Gene 1.0 ST Array 10805670 - -
          Affymetrix Rat Gene 1.0 ST Array 10805656 - -
          Affymetrix Rat Gene 1.0 ST Array 10805654 - -
          Affymetrix Rat Gene 1.0 ST Array 10805664 - -
          Affymetrix Rat Gene 1.0 ST Array 10805666 - -
          Affymetrix Rat Gene 1.0 ST Array 10805659 - -
          Affymetrix Rat Gene 1.0 ST Array 10805655 - -
          Affymetrix Rat Gene 1.0 ST Array 10805663 - -
          Affymetrix Rat Gene 1.0 ST Array 10805657 - -
          Affymetrix Rat Gene 1.0 ST Array 10805653 - -
          Affymetrix Rat Gene 1.0 ST Array 10805661 - -
          Affymetrix Rat Gene 1.0 ST Array 10805660 - -
          Affymetrix Rat Gene 1.1 ST Array 10805652 - -
          Rat Genome 230 2.0 Array 1390822_at - -
          Rat Genome 230 2.0 Array 1390822_PM_at - -
          Affymetrix HT Rat Focus Array 1390822_at - -
          Affymetrix HT Rat Focus Array 1390822_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900000154.rn.1 TC1900000015.rn.1 RTA TC
          Rat Expression Set 230 1390822_PM_at - -
          Rat Expression Set 230 1390822_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805652 - -
          Rat Genome U34 Set rc_AA891858_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22324225 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline