Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536855_m1
          See other POLR2C GE Assays ›
          Gene Symbol:
          Polr2c
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide C
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 10575060 - 10581842 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536855_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012473.1 NP_001012491.1 2 - 3 161 75
          GenBank mRNA BC089905.1 - 2 - 3 161 7322752 75
          FQ218345.1 - 2 - 3 78 75
          FQ226282.1 - 2 - 3 200 75
          FQ227427.1 - 2 - 3 181 75
          FQ235323.1 - 2 - 3 200 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000987.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Polr2c

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          RNA polymerase II activity
          DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P321329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395422 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12296 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321329 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P321329 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P321329 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P321329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809250 - -
          Affymetrix Rat Gene 1.0 ST Array 10809245 - -
          Affymetrix Rat Gene 1.0 ST Array 10809246 - -
          Affymetrix Rat Gene 1.0 ST Array 10809248 - -
          Affymetrix Rat Gene 1.0 ST Array 10809243 - -
          Affymetrix Rat Gene 1.0 ST Array 10809247 - -
          Affymetrix Rat Gene 1.0 ST Array 10809242 - -
          Affymetrix Rat Gene 1.0 ST Array 10809249 - -
          Affymetrix Rat Gene 1.0 ST Array 10809244 - -
          Affymetrix Rat Gene 1.1 ST Array 10809240 - -
          Rat Genome 230 2.0 Array 1398899_at - -
          Rat Genome 230 2.0 Array 1390491_PM_at - -
          Rat Genome 230 2.0 Array 1388570_PM_at - -
          Rat Genome 230 2.0 Array 1372300_at - -
          Rat Genome 230 2.0 Array 1398899_PM_at - -
          Rat Genome 230 2.0 Array 1390491_at - -
          Rat Genome 230 2.0 Array 1372300_PM_at - -
          Rat Genome 230 2.0 Array 1388570_at - -
          Affymetrix HT Rat Focus Array 1398899_at - -
          Affymetrix HT Rat Focus Array 1398899_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900007875.rn.1 TC1900000987.rn.1 RTA TC
          Rat Expression Set 230 1398899_PM_at - -
          Rat Expression Set 230 1398899_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809240 - -
          Rat Genome U34 Set rc_AI170414_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22009757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0