Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01640240_m1
          See other XIRP2 GE Assays ›
          Gene Symbol:
          Xirp2
          Gene Name:
          xin actin-binding repeat containing 2
          Gene Aliases:
          Cmya3, LACS
          Chromosome Location:
          Chr.3: 53563245 - 53658113 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01640240_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xirp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201989.1 NP_973718.1 7 - 8 10047 109
          GenBank mRNA AF466694.1 - 7 - 8 10047 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000721.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xirp2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ventricular septum development
          heart development
          actin cytoskeleton organization
          cell-cell junction organization
          cardiac muscle tissue morphogenesis
          actin binding
          alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P231223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12368 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160399 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123105 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231223 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P231223 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P231223 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P231223 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71383 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91844 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66591 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836507 - -
          Affymetrix Rat Gene 1.0 ST Array 10836506 - -
          Affymetrix Rat Gene 1.0 ST Array 10836513 - -
          Affymetrix Rat Gene 1.0 ST Array 10836510 - -
          Affymetrix Rat Gene 1.0 ST Array 10836508 - -
          Affymetrix Rat Gene 1.0 ST Array 10836512 - -
          Affymetrix Rat Gene 1.0 ST Array 10836514 - -
          Affymetrix Rat Gene 1.0 ST Array 10836505 - -
          Affymetrix Rat Gene 1.0 ST Array 10836511 - -
          Affymetrix Rat Gene 1.0 ST Array 10836509 - -
          Affymetrix Rat Gene 1.1 ST Array 10836504 - -
          Affymetrix Rat Clariom S Assay PSR0300005384.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005381.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005382.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005383.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005387.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005385.rn.1 TC0300000721.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10836504 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20927247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline