Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747967_m1
          See other POT1 GE Assays ›
          Gene Symbol:
          Pot1
          Gene Name:
          protection of telomeres 1
          Gene Aliases:
          Pot1a
          Chromosome Location:
          Chr.4: 51888632 - 51946764 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pot1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024322.1 NP_001019493.1 16 - 17 1994 96
          XM_006236160.2 XP_006236222.1 16 - 17 2012 96
          XM_006236161.2 XP_006236223.1 17 - 18 2234 96
          XM_008762809.1 XP_008761031.1 17 - 18 2416 96
          XM_008762810.1 XP_008761032.1 17 - 18 2361 96
          XM_008762811.1 XP_008761033.1 15 - 16 1965 96
          GenBank mRNA BC085834.1 - 16 - 17 1994 7134840 96
          BC090024.1 - 10 - 11 1336 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002552.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pot1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          telomere maintenance via telomerase
          telomere capping
          telomere assembly
          negative regulation of telomere maintenance via telomerase
          positive regulation of telomere maintenance via telomerase
          DNA duplex unwinding
          positive regulation of helicase activity
          chromosome organization
          positive regulation of telomerase activity
          negative regulation of telomerase activity
          positive regulation of DNA strand elongation
          telomerase inhibitor activity
          DEAD/H-box RNA helicase binding
          single-stranded telomeric DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P16623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51180 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170825 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135293 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P16623 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P16623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85562 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47232 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63639 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72728 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10861370 - -
          Affymetrix Rat Gene 1.0 ST Array 10861372 - -
          Affymetrix Rat Gene 1.0 ST Array 10861375 - -
          Affymetrix Rat Gene 1.0 ST Array 10861368 - -
          Affymetrix Rat Gene 1.0 ST Array 10861362 - -
          Affymetrix Rat Gene 1.0 ST Array 10861367 - -
          Affymetrix Rat Gene 1.0 ST Array 10861371 - -
          Affymetrix Rat Gene 1.0 ST Array 10861376 - -
          Affymetrix Rat Gene 1.0 ST Array 10861364 - -
          Affymetrix Rat Gene 1.0 ST Array 10861369 - -
          Affymetrix Rat Gene 1.0 ST Array 10861377 - -
          Affymetrix Rat Gene 1.0 ST Array 10861366 - -
          Affymetrix Rat Gene 1.0 ST Array 10861365 - -
          Affymetrix Rat Gene 1.0 ST Array 10861374 - -
          Affymetrix Rat Gene 1.0 ST Array 10861363 - -
          Affymetrix Rat Gene 1.0 ST Array 10861378 - -
          Affymetrix Rat Gene 1.0 ST Array 10861373 - -
          Affymetrix Rat Gene 1.1 ST Array 10861361 - -
          Rat Genome 230 2.0 Array 1399134_at - -
          Rat Genome 230 2.0 Array 1386777_PM_at - -
          Rat Genome 230 2.0 Array 1386777_at - -
          Rat Genome 230 2.0 Array 1399134_PM_at - -
          Affymetrix HT Rat Focus Array 1386777_PM_at - -
          Affymetrix HT Rat Focus Array 1399134_at - -
          Affymetrix HT Rat Focus Array 1386777_at - -
          Affymetrix HT Rat Focus Array 1399134_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400018089.rn.1 TC0400002552.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400018086.rn.1 TC0400002552.rn.1 RTA TC
          Rat Expression Set 230 1399134_PM_at - -
          Rat Expression Set 230 1386777_at - -
          Rat Expression Set 230 1386777_PM_at - -
          Rat Expression Set 230 1399134_at - -
          Affymetrix Rat Gene 1.0 ST Array 10861361 - -
          Rat Genome U34 Set rc_AI030751_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22284874 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373773 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline