Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01748691_m1
          See other POLRMT GE Assays ›
          Gene Symbol:
          Polrmt
          Gene Name:
          polymerase (RNA) mitochondrial (DNA directed)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 12840912 - 12851153 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01748691_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polrmt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106766.1 NP_001100236.1 3 - 4 758 82
          XM_006240859.2 XP_006240921.1 3 - 4 784 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000231.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from mitochondrial promoter
          DNA binding
          DNA-directed RNA polymerase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38617 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123969 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123969 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31961 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894047 - -
          Affymetrix Rat Gene 1.0 ST Array 10894049 - -
          Affymetrix Rat Gene 1.0 ST Array 10894043 - -
          Affymetrix Rat Gene 1.0 ST Array 10894031 - -
          Affymetrix Rat Gene 1.0 ST Array 10894038 - -
          Affymetrix Rat Gene 1.0 ST Array 10894040 - -
          Affymetrix Rat Gene 1.0 ST Array 10894039 - -
          Affymetrix Rat Gene 1.0 ST Array 10894032 - -
          Affymetrix Rat Gene 1.0 ST Array 10894036 - -
          Affymetrix Rat Gene 1.0 ST Array 10894035 - -
          Affymetrix Rat Gene 1.0 ST Array 10894045 - -
          Affymetrix Rat Gene 1.0 ST Array 10894033 - -
          Affymetrix Rat Gene 1.0 ST Array 10894034 - -
          Affymetrix Rat Gene 1.0 ST Array 10894029 - -
          Affymetrix Rat Gene 1.0 ST Array 10894041 - -
          Affymetrix Rat Gene 1.0 ST Array 10894046 - -
          Affymetrix Rat Gene 1.0 ST Array 10894030 - -
          Affymetrix Rat Gene 1.0 ST Array 10894044 - -
          Affymetrix Rat Gene 1.0 ST Array 10894037 - -
          Affymetrix Rat Gene 1.0 ST Array 10894048 - -
          Affymetrix Rat Gene 1.0 ST Array 10894042 - -
          Affymetrix Rat Gene 1.1 ST Array 10894028 - -
          Rat Genome 230 2.0 Array 1374295_PM_at - -
          Rat Genome 230 2.0 Array 1374295_at - -
          Affymetrix HT Rat Focus Array 1374295_at - -
          Affymetrix HT Rat Focus Array 1374295_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700002211.rn.1 TC0700000231.rn.1 RTA TC
          Rat Expression Set 230 1374295_PM_at - -
          Rat Expression Set 230 1374295_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894028 - -
          Rat Genome U34 Set rc_AI233096_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline