Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01749582_m1
          See other DHRS3 GE Assays ›
          Gene Symbol:
          Dhrs3
          Gene Name:
          dehydrogenase/reductase (SDR family) member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 162809090 - 162843385 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01749582_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhrs3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037199.3 NP_001032276.3 4 - 5 996 77
          GenBank mRNA BC107450.1 - 4 - 5 996 7322355 77
          EF125189.1 - 4 - 5 1442 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001873.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhrs3

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          outflow tract morphogenesis
          regulation of ossification
          negative regulation of retinoic acid receptor signaling pathway
          oxidation-reduction process
          palate development
          bone morphogenesis
          cardiac septum morphogenesis
          retinol dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P621633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145245 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P621633 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P621633 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P621633 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P621633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26434 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77086 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10873840 - -
          Affymetrix Rat Gene 1.0 ST Array 10873843 - -
          Affymetrix Rat Gene 1.0 ST Array 10873849 - -
          Affymetrix Rat Gene 1.0 ST Array 10873842 - -
          Affymetrix Rat Gene 1.0 ST Array 10873847 - -
          Affymetrix Rat Gene 1.0 ST Array 10873841 - -
          Affymetrix Rat Gene 1.0 ST Array 10873848 - -
          Affymetrix Rat Gene 1.0 ST Array 10873846 - -
          Affymetrix Rat Gene 1.0 ST Array 10873845 - -
          Affymetrix Rat Gene 1.0 ST Array 10873839 - -
          Affymetrix Rat Gene 1.1 ST Array 10873838 - -
          Rat Genome 230 2.0 Array 1373240_at - -
          Rat Genome 230 2.0 Array 1373240_PM_at - -
          Affymetrix HT Rat Focus Array 1373240_at - -
          Affymetrix HT Rat Focus Array 1373240_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500014030.rn.1 TC0500001873.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014031.rn.1 TC0500001873.rn.1 RTA TC
          Rat Expression Set 230 1373240_at - -
          Rat Expression Set 230 1373240_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10873838 - -
          Rat Genome U34 Set rc_AA965122_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21142883 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline