Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750102_m1
          See other ZFP583 GE Assays ›
          Gene Symbol:
          Zfp583
          Gene Name:
          zinc finger protein 583
          Gene Aliases:
          RGD1562044
          Chromosome Location:
          Chr.1: 71244094 - 71262774 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750102_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp583
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134609.2 NP_001128081.2 4 - 5 425 118
          XM_006228197.2 XP_006228259.1 4 - 5 618 118
          XM_006228199.2 XP_006228261.1 4 - 5 615 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000818.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P728540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260834 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P728540 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P728540 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P728540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10703867 - -
          Affymetrix Rat Gene 1.0 ST Array 10703865 - -
          Affymetrix Rat Gene 1.0 ST Array 10703866 - -
          Affymetrix Rat Gene 1.1 ST Array 10703864 - -
          Affymetrix Rat Clariom S Assay PSR0100005365.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005368.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005356.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005353.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005359.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005364.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005361.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005366.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005370.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005373.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005371.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005369.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005352.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100005363.rn.1 TC0100000818.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10703864 - -
          Rat Genome U34 Set rc_AA900483_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21390946 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360210 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline