Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753182_m1
          See other SPAG6 GE Assays ›
          Gene Symbol:
          Spag6
          Gene Name:
          sperm associated antigen 6
          Gene Aliases:
          RGD1310892
          Chromosome Location:
          Chr.11: 88424493 - 88517910 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753182_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spag6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034960.1 NP_001030132.1 10 1512 76
          GenBank mRNA BC104678.1 - 10 1512 7454069 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001031.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spag6

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s)

          spermatogenesis
          sperm motility

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18519 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902818 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111779 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47297 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094910 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094910 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P293830 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51136 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98221 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77184 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21660 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78481 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63240 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752567 - -
          Affymetrix Rat Gene 1.0 ST Array 10752573 - -
          Affymetrix Rat Gene 1.0 ST Array 10752571 - -
          Affymetrix Rat Gene 1.0 ST Array 10752565 - -
          Affymetrix Rat Gene 1.0 ST Array 10752569 - -
          Affymetrix Rat Gene 1.0 ST Array 10752564 - -
          Affymetrix Rat Gene 1.0 ST Array 10752566 - -
          Affymetrix Rat Gene 1.0 ST Array 10752570 - -
          Affymetrix Rat Gene 1.0 ST Array 10752568 - -
          Affymetrix Rat Gene 1.0 ST Array 10752572 - -
          Affymetrix Rat Gene 1.1 ST Array 10752563 - -
          Affymetrix Rat Clariom S Assay PSR1100006205.rn.1 TC1100001031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006206.rn.1 TC1100001031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006208.rn.1 TC1100001031.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10752563 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22072315 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline