Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01753863_m1
          See other ARL5A GE Assays ›
          Gene Symbol:
          Arl5a
          Gene Name:
          ADP-ribosylation factor-like 5A
          Gene Aliases:
          Arl5
          Chromosome Location:
          Chr.3: 37922544 - 37947434 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01753863_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053979.2 NP_446431.1 5 - 6 685 80
          XM_008761696.1 XP_008759918.1 5 - 6 757 80
          GenBank mRNA BC097294.1 - 5 - 6 685 7458249 80
          X78604.1 - 5 - 6 534 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003110.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl5a

          Panther Classification:

          Molecular Function -

          G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P159360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37329 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93854 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159360 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159360 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P159360 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14186 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700199826 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700821351 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81438 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63554 - -
          Agilent Rat cDNA Microarray (G4105A) 700821351 - -
          Agilent Rat cDNA Microarray (G4105A) 700199826 - -
          Agilent Rat cDNA Microarray (G4105A) 700504436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845303 - -
          Affymetrix Rat Gene 1.0 ST Array 10845301 - -
          Affymetrix Rat Gene 1.0 ST Array 10845304 - -
          Affymetrix Rat Gene 1.0 ST Array 10845302 - -
          Affymetrix Rat Gene 1.0 ST Array 10845300 - -
          Affymetrix Rat Gene 1.0 ST Array 10845299 - -
          Affymetrix Rat Gene 1.0 ST Array 10845305 - -
          Affymetrix Rat Gene 1.1 ST Array 10845298 - -
          Rat Genome 230 2.0 Array 1369729_PM_at - -
          Rat Genome 230 2.0 Array 1374178_at - -
          Rat Genome 230 2.0 Array 1374178_PM_at - -
          Rat Genome 230 2.0 Array 1369729_at - -
          Affymetrix HT Rat Focus Array 1369729_PM_at - -
          Affymetrix HT Rat Focus Array 1369729_at - -
          Affymetrix Rat Clariom S Assay PSR0300024129.rn.1 TC0300003110.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300024128.rn.1 TC0300003110.rn.1 RTA TC
          Rat Expression Set 230 1369729_PM_at - -
          Rat Expression Set 230 1369729_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845298 - -
          Rat Genome U34 Set rc_AA956958_at - -
          Rat Genome U34 Set rc_AA875135_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22273383 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360968 - -
          RatRef-12 Expression BeadChip ILMN_61137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline