Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754303_m1
          See other NKAIN4 GE Assays ›
          Gene Symbol:
          Nkain4
          Gene Name:
          Na+/K+ transporting ATPase interacting 4
          Gene Aliases:
          RGD1305809
          Chromosome Location:
          Chr.3: 176444545 - 176465197 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754303_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkain4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106550.1 NP_001100020.1 4 - 5 469 55
          XM_006235744.2 XP_006235806.1 6 - 7 723 55
          XM_006235745.2 XP_006235807.1 6 - 7 2605 55
          XM_008762496.1 XP_008760718.1 7 - 8 1072 55
          XM_008762497.1 XP_008760719.1 7 - 8 1900 55
          XM_008762498.1 XP_008760720.1 7 - 8 875 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005033.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55236 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000281 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000281 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000281 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P148416 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342520 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92297 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1037 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852422 - -
          Affymetrix Rat Gene 1.0 ST Array 10852427 - -
          Affymetrix Rat Gene 1.0 ST Array 10852424 - -
          Affymetrix Rat Gene 1.0 ST Array 10852423 - -
          Affymetrix Rat Gene 1.1 ST Array 10852421 - -
          Rat Genome 230 2.0 Array 1390037_at - -
          Rat Genome 230 2.0 Array 1390037_PM_at - -
          Affymetrix HT Rat Focus Array 1390037_at - -
          Affymetrix HT Rat Focus Array 1390037_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300038257.rn.1 TC0300005033.rn.1 RTA TC
          Rat Expression Set 230 1390037_PM_at - -
          Rat Expression Set 230 1390037_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852421 - -
          Rat Genome U34 Set rc_AI102026_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21118831 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline