Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01771406_g1
          See other IGHG GE Assays ›
          Gene Symbol:
          Ighg
          Gene Name:
          Immunoglobulin heavy chain (gamma polypeptide)
          Gene Aliases:
          RGD1359539
          Chromosome Location:
          Chr.6: 139138229 - 139158896 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01771406_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ighg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC088271.1 - 754 7379698 89
          BC092584.1 - 772 7384581 89
          BC095846.1 - 758 7388075 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095554 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P487269 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892625 - -
          Affymetrix Rat Gene 1.0 ST Array 10887658 - -
          Affymetrix Rat Gene 1.0 ST Array 10892513 - -
          Affymetrix Rat Gene 1.0 ST Array 10892422 - -
          Affymetrix Rat Gene 1.0 ST Array 10892424 - -
          Affymetrix Rat Gene 1.0 ST Array 10892540 - -
          Affymetrix Rat Gene 1.0 ST Array 10887649 - -
          Affymetrix Rat Gene 1.0 ST Array 10892504 - -
          Affymetrix Rat Gene 1.0 ST Array 10892423 - -
          Affymetrix Rat Gene 1.0 ST Array 10887654 - -
          Affymetrix Rat Gene 1.0 ST Array 10892505 - -
          Affymetrix Rat Gene 1.0 ST Array 10892442 - -
          Affymetrix Rat Gene 1.0 ST Array 10887659 - -
          Affymetrix Rat Gene 1.0 ST Array 10892514 - -
          Affymetrix Rat Gene 1.0 ST Array 10887648 - -
          Affymetrix Rat Gene 1.0 ST Array 10892539 - -
          Affymetrix Rat Gene 1.1 ST Array 10892509 - -
          Affymetrix Rat Gene 1.1 ST Array 10892409 - -
          Affymetrix Rat Gene 1.1 ST Array 10892536 - -
          Affymetrix Rat Gene 1.1 ST Array 10887657 - -
          Affymetrix Rat Gene 1.1 ST Array 10892620 - -
          Affymetrix Rat Clariom S Assay PSR0600023102.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023176.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023211.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023156.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023196.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023101.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023178.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023100.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600023103.rn.1 TC0600003269.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10892409 - -
          Affymetrix Rat Gene 1.0 ST Array 10892620 - -
          Rat Genome U34 Set rc_AA893565_at - -
          Rat Genome U34 Set L07401_f_at - -
          Rat Genome U34 Set M28670_at - -
          Rat Genome U34 Set L07402_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22080253 - -
          ABI Rat Genome Survey Microarray 22175405 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355545 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline