Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01775382_m1
          See other LOC291863 GE Assays ›
          Gene Symbol:
          LOC291863
          Gene Name:
          carboxylesterase-like
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01775382_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC291863
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013889.1 NP_001013911.1 11 - 12 1190 110
          GenBank mRNA BC078681.1 - 11 - 12 1190 7123712 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000150.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC291863

          Panther Classification:

          Molecular Function -

          esterase

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P555689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P555689 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P555689 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555689 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81562 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38678 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931277 - -
          Affymetrix Rat Gene 1.0 ST Array 10931281 - -
          Affymetrix Rat Gene 1.0 ST Array 10931280 - -
          Affymetrix Rat Gene 1.0 ST Array 10931272 - -
          Affymetrix Rat Gene 1.0 ST Array 10931284 - -
          Affymetrix Rat Gene 1.0 ST Array 10931273 - -
          Affymetrix Rat Gene 1.0 ST Array 10931275 - -
          Affymetrix Rat Gene 1.0 ST Array 10931278 - -
          Affymetrix Rat Gene 1.0 ST Array 10931283 - -
          Affymetrix Rat Gene 1.0 ST Array 10931279 - -
          Affymetrix Rat Gene 1.0 ST Array 10931282 - -
          Affymetrix Rat Gene 1.0 ST Array 10931274 - -
          Affymetrix Rat Gene 1.0 ST Array 10931276 - -
          Affymetrix Rat Gene 1.1 ST Array 10931271 - -
          Affymetrix Rat Gene 1.1 ST Array 10806221 - -
          Affymetrix Rat Clariom S Assay PSR1900001245.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001253.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001249.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001250.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001252.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001254.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001260.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001213.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001255.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001257.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001247.rn.1 TC1900000150.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10931271 - -
          Rat Genome U34 Set rc_AA800851_i_at - -
          Rat Genome U34 Set rc_AA800851_r_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649844 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline